MicroAmp™ Cap Installing Tool
Applied Biosystems™

MicroAmp™ Cap Installing Tool

The MicroAmp™ Cap Installing Tool helps you install strips of MicroAmp™ caps onto MicroAmp™ reaction tubes in a tray⁄retainer setRead more
Have Questions?
Catalog NumberQuantity
43300151 Tool
Catalog number 4330015
Price (EUR)
67,25
Each
Quantity:
1 Tool
Price (EUR)
67,25
Each
The MicroAmp™ Cap Installing Tool helps you install strips of MicroAmp™ caps onto MicroAmp™ reaction tubes in a tray⁄retainer set or 96-well reaction plate.

• Available in handle style
• Seal 8 or 12 cap strips
• Recessed cup for pressing caps onto tubes
• Convenient tool ensures proper sealing

Press and Seal Caps
The MicroAmp™ Cap Installing Tool enables you to ergonomically seal MicroAmp™ reaction tubes and plates. This tight seal will prevent well-to-well contamination and sample loss.

Used With
MicroAmp™ Optical 8-Cap Strip, MicroAmp™ Optical 8-Tube Strip, 0.2 ml, MicroAmp™ Optical 96-Well Reaction Plate with Barcode, MicroAmp™ Optical 96-Well Reaction Plate with Barcode.

Note: For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)2720 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOne™, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, 7500 Fast Dx System, 2720 Thermal Cycler, ViiA™ 7 Fast System, QuantStudio™ Dx
Product LineMicroAmp
Product TypeCap Installing Tool
Quantity1 Tool
Shipping ConditionRoom Temperature
Format12-cap Strip, 8-cap Strip
Unit SizeEach
Contents & Storage
Cap installing tool for MicroAmp plates, tubes, and caps