Product Image
Applied Biosystems™
GeneMapper™ Software v4.1, full installation
GeneMapper™ Software is a flexible fragment analysis software package that provides quality DNA sizing and allele calls for all Applied Biosystems™ genetic analyzers.
Have Questions?
Catalog NumberQuantity
43669251 License
Catalog number 4366925
Price (USD)
***
Quantity:
1 License
Price (USD)
***
GeneMapper™ Software is a flexible fragment analysis software package that provides quality DNA sizing and allele calls for all Applied Biosystems™ genetic analyzers. This software is designed to support multi-application functionality, including analysis of amplified fragment length polymorphism (AFLP), loss of heterozygosity (LOH), microsatellites, and SNP genotyping.
Features include:
  • Multi-user, client-server deployment (through Oracle 11g RDBMS)
  • Increased data processing efficiency through remote auto-analysis and command line operation
  • Security and audit features to help you meet 21CFR11 requirements
  • Organize, manage, and batch-process your data with Report Manager and Database Dashboard
  • Automatically evaluate and reduce data review time with Process Quality Values (PQVs) for high-throughput genotyping
The software supports a wide variety of applications on the Applied Biosystems™ 3500, 3500xL, 3730, 3730xl, 3130, 3130xl, 3100, 3100–Avant, and 310 instrument systems.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntryGenetic Analyzer Data
Data OutputsSize, Genotype, and Process Quality (SQ, GQ, PQV) Reports, Sample and Genotype Plots
For Use With (Application)Sequencing
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3700 Genetic Analyzer, 3500 Dx Genetic Analyzer, 3730xl DNA Analyzer, 3130 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
LicenseClient, Server
Operating SystemWindows XP Pro, Windows Vista Business, Windows™ XP Pro SP2, Windows™ Vista Bus SP1 32-bit, Windows™ XP Pro SP3
Product LineGeneMapper
Product TypeSoftware
Quantity1 License
Shipping ConditionRoom Temperature
Software1 Full License, GeneMapper™, Genotyping
System RequirementsComputer: Core 2 Duo E8400/3 GHz; 2 GB RAM
Unit Size1 license,1 copy
Contents & Storage
Software/CD
Frequently asked questions (FAQs)
How can I import Project Information into GeneMapper Manager?
Go to the Tools menu and select GeneMapper Manager. Click on the tab for the file(s) you want to import. Click on Import at the bottom of the window and select the files to be imported. Note: Multiple projects can be selected for importing at one time, but all other tabs are one at a time. Once imported, the new files will appear in the list. When finished, click Done in the lower right corner.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up my data files for DNA sequencing and fragment analysis applications, with the following extension, .ab1 or .fsa, on the capillary electrophoresis instruments?
The location of the sample files, .ab1 or .fsa, will either be in the Runs folder (D:/AppliedBio/310/Runs) on the ABI PRISM 310 Genetic Analyzer system or it will be determined by the Results Group in the Data Collection Software on the multi-capillary platforms. If you do not know where that location is, you can go to the Data Collection Software for your instrument and go to the Results Group section (top left corner in Unified Data Collection; for 3500 Data Collection, go to Library and in the Manage section, select Results Group), open the Results Group, and check the Sample File Folder location.
Save the files to the storage media of your choice (CD, DVD, network drive, flash drive).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Do you have a manual uninstall procedure for Full Install configurations on a stand-alone computer for GeneMapper Software v.4.1?
Please download this PDF (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v.4-uninstall-full-install.pdf) and follow the directions to uninstall GeneMapper Software v.4.1.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
I need to uninstall GeneMapper Software v.4.1 Client because of failed installation. How can I do this for Client Install configurations on a stand-alone computer?
Please download this PDF (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v.4-1-uninstall-client-install.pdf) and follow the directions to uninstall GeneMapper Software v.4.1.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We are upgrading to the Windows 7 operating system. Can I install GeneMapper Software v.4.1 on that?
Unfortunately, GeneMapper Software v.4.1 is not compatible with Windows 7. Please click here (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v.4-1-datasheet.pdf) for the GeneMapper Software v.4.1 datasheet.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.