Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
Product Image
Applied Biosystems™

TaqMan™ Array Mouse GPCR Panel

The Applied Biosystems™ TaqMan™ Mouse GPCR Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis ofRead more
Have Questions?
Catalog NumberQuantity
43787034 cards
Catalog number 4378703
Price (USD)
2,635.00
Each
Quantity:
4 cards
Price (USD)
2,635.00
Each
The Applied Biosystems™ TaqMan™ Mouse GPCR Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of mouse GPCR genes important in drug discovery. This product is designed with 380 TaqMan™ Gene Expression Assays arrayed in singletons in 384 wells (plus 1 control arrayed in quadruplicate) and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. The targets include retinal receptors, small molecule receptors, chemokine receptors and others including classic endogenous control targets. This product is packaged in 1 box containing 4 cards.

The GPCR targets selected fall into the 50 subfamilies listed, many of which are drug targets or disease associated. Odorant, olfactory, gustatory or pheromone receptors are not included on these arrays. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 2 μL Reactions
Product LineTaqMan
Quantity4 cards
Research CategoryCell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Reproduction, Toxicology & Drug Metabolism, Immunology, Development
SpeciesMouse
Target50 GPCR subfamilies (excludes odorant, olfactory, gustatory and pheromone receptors)
For Use With (Application)Gene Expression
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.