This software includes an upgrade for the 7300 SDS Software to provide the capability and reliability of the most recentRead more
Have Questions?
Catalog Number
Quantity
4379635
1 kit
Catalog number 4379635
Price (USD)
508.00
Each
Quantity:
1 kit
Price (USD)
508.00
Each
This software includes an upgrade for the 7300 SDS Software to provide the capability and reliability of the most recent version. Powerful and user-friendly, the 7300 Sequence Detection Software includes: • Simple plate setup wizard is streamlined for each application – option to setup before or after the completing the run • Correct plate-setup mistakes after the run without losing collected data • One-click graphical export to PowerPoint or JPG for top notch presentations • Flexible system can be used as a plate reader or a regular thermal cycler if needed • Real-time amplification monitoring and flexibility to add additional PCR cycles during a run • Lamp-lifetime monitoring ensures highest possible instrument performance
Additionally, the 7300 System SDS RQ Study Software, a gene expression analysis software package with powerful data viewing capabilities, is included. Data from up to 10 96-well RQ plates can be simultaneously analyzed. In an RQ study, you can: • Select the endogenous control and the calibrator sample. • Select the control type when applicable. • Set baseline and threshold values and RQ Min/Max Confidence Levels. • Omit individual wells or sample replicates.
The user is able to view post-run amplification in three different formats, while gene expression plots show expression levels of target samples relative to the calibrator.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)7300 System
Operating SystemWindows XP Pro (Service Pk. 2)
Quantity1 kit
Software TypeGene Expression Analysis
Product TypeSoftware Upgrade Kit
Unit SizeEach
Contents & Storage
Store at Room Temperature
Citations & References (1145)
Citations & References
Abstract
Authors:
Journal:
PubMed ID:11282963
ADP-ribosylation factor-like GTPase ARFRP1 is required for trans-Golgi to plasma membrane trafficking of E-cadherin.
Authors:Zahn C, Jaschke A, Weiske J, Hommel A, Hesse D, Augustin R, Lu L, Hong W, Florian S, Scheepers A, Joost HG, Huber O, Schurmann A
Journal:J Biol Chem
PubMed ID:18662990
ADP-ribosylation factor-related protein 1 (ARFRP1) plays a specific role in Golgi function controlling recruitment of GRIP domain proteins and ARL1 to the trans-Golgi. Deletion of the mouse Arfrp1 gene causes embryonic lethality during early gastrulation, because epiblast cells detach from the ectodermal cell layer and do not differentiate to mesodermal ... More
Preclinical pharmacokinetic and pharmacodynamic evaluation of metronomic and conventional temozolomide dosing regimens.
Authors:Zhou Q, Guo P, Wang X, Nuthalapati S, Gallo JM
Journal:J Pharmacol Exp Ther
PubMed ID:17259446
Metronomic dosed (MD) chemotherapy as opposed to conventional dosed (CD) chemotherapy is considered an alternate strategy to target angiogenesis and limit host toxicity. Although this approach is promising, there has not been any attempt to define optimal metronomic dosing regimens by integrating pharmacokinetic (PK) with pharmacodynamic (PD) measurements. The aim ... More
Aim2 Deficiency Stimulates the Expression of IFN-Inducible Ifi202, a Lupus Susceptibility Murine Gene within the Nba2 Autoimmune Susceptibility Locus.
Authors:Panchanathan R, Duan X, Shen H, Rathinam VA, Erickson LD, Fitzgerald KA, Choubey D
Journal:J Immunol
PubMed ID:21057088
Murine Aim2 and p202 proteins (encoded by the Aim2 and Ifi202 genes) are members of the IFN-inducible p200 protein family. Both proteins can sense dsDNA in the cytoplasm. However, upon sensing dsDNA, only the Aim2 protein through its pyrin domain can form an inflammasome to activate caspase-1 and induce cell ... More
Small RNA class transition from siRNA/piRNA to miRNA during pre-implantation mouse development.
Authors:Ohnishi Y, Totoki Y, Toyoda A, Watanabe T, Yamamoto Y, Tokunaga K, Sakaki Y, Sasaki H, Hohjoh H
Journal:Nucleic Acids Res
PubMed ID:20385573
Recent studies showed that small interfering RNAs (siRNAs) and Piwi-interacting RNA (piRNA) in mammalian germ cells play important roles in retrotransposon silencing and gametogenesis. However, subsequent contribution of those small RNAs to early mammalian development remains poorly understood. We investigated the expression profiles of small RNAs in mouse metaphase II ... More