VetMAX™-Gold BVDV PI Detection Kit
Applied Biosystems™

VetMAX™-Gold BVDV PI Detection Kit

The Bovine Virus Diarrhea RNA Test Kit (VetMAX™-Gold BVDV PI Detection Kit) is intended for use in the rapid, inRead more
Have Questions?
Catalog NumberQuantity
4413938100 reactions
Catalog number 4413938
Price (MXN)
-
Quantity:
100 reactions
The Bovine Virus Diarrhea RNA Test Kit (VetMAX™-Gold BVDV PI Detection Kit) is intended for use in the rapid, in vitro detection of bovine virus diarrhea virus (BVDV RNA) extracted from ear notches in acute and persistently infected (PI) cattle.

The VetMAX™-Gold BVDV PI Detection Kit is the first USDA-licensed real-time PCR test for the detection of BVDV in cattle. The assay is a single-tube, real-time, reverse transcription-polymerase chain reaction (RT-PCR) in which RNA is reverse transcribed into cDNA, and BVDV targets are amplified and detected in real time using fluorescent TaqMan™ probes.

Have Confidence in Your Results:
• Proven repeatability and reproducibility in field studies
• Highly specific and sensitive detection of type 1 and type 2 BVDV
• Consistently detects <40 RNA copies
• Contains Xeno™ RNA internal positive control for confirmation of results
• Rapid and cost-effective method for detection of BVDV with results in ∼1.5 hours
• Simple, fast workflow minimizes sample handling and reduces sample cross-contamination
• Easily automated for high-throughput processing
• Detects BVDV in ear-notch tissue
• Robust and reliable AmpliTaq Gold™ DNA Polymerase and TaqMan™ technology

Limitations:
• The kit is not intended for discriminating viral subtypes
• Samples should be handled as recommended to prevent degradation of any BVDV RNA that is present
• RNA extraction methods should yield RNA free of RT-PCR inhibitors, which can prevent amplification of target RNA
• Follow “good PCR practices” to prevent false positive amplifications due to contamination of test samples with PCR products
For Veterinary Use Only
Specifications
FormatTube
No. of Reactions100 Reactions
PolymeraseDNA Polymerase
Product LineVetMAX
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesCattle/Bovine
Target Organism ClassBovine Viral Diarrhea Virus (BVDV)
Detection MethodPrimer-probe
For Use With (Application)Pathogen Detection
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C. Includes:
RT-PCR Enzyme Mix
RT-PCR Buffer
BVDV Primer Probe Mix
Xeno™ RNA Control
BVDV Control RNA
Nucleic Acid Dilution Solution

Store at -30°C to 25°C:
Nuclease-free Water

Frequently asked questions (FAQs)

With the VetMAX-Gold BVDV PI Detection Kit, I am getting signal detection in the no-template control (NTC) reaction. Why is this?

This is likely due to PCR contamination. Here are some recommendations:

- Repeat the qRT-PCR reaction with fresh reagents and freshly decontaminated pipettors.
- Set up and run the qRT-PCR reaction in an area that is isolated from areas used for RNA purification and PCR product analysis.