3500xL Genetic Analyzer for Resequencing & Fragment Analysis
Applied Biosystems™

3500xL Genetic Analyzer for Resequencing & Fragment Analysis

The 3500xL 24-capillary platform can run a wide variety of applications, including de novo sequencing and resequencing (mutational profiling), asMás información
Have Questions?
Número de catálogoCantidad
44404631 sistema
Número de catálogo 4440463
Precio (MXN)
-
Cantidad:
1 sistema
The 3500xL 24-capillary platform can run a wide variety of applications, including de novo sequencing and resequencing (mutational profiling), as well as microsatellite analysis, MLPA™, LOH, MLST, AFLP™ and SNP validation or screening. The majority of applications can be run on a single polymer and capillary array.

This package includes a 24-capillary instrument, 3500 Series Data Collection Software, a Dell Workstation and monitor, as well as reagent kits for system qualification and secondary analysis software. The 3500 Series Data Collection Software supports sequencing and fragment analysis, and integrates seamlessly with downstream software for secondary analysis of genetic data. This package includes such secondary software: GeneMapper™ software for fragment analysis and software for resequencing applications.

Features include:

• An 8 capillary system that can easily be upgraded to a 24-capillary system when you’re ready.
• Single-line, 505 nm, solid-state, long-life laser that utilizes a standard power supply and requires no heat-removal ducting
• Powerful, integrated data collection and primary analysis software provides real-time assessment of data quality
• Radio frequency identification (RFID) technology tracks key consumables data and records administrative information
• Advanced multiplexing capabilities for DNA fragment analysis with up to six unique dyes
• Unrivaled application flexibility—one array and one polymer are used for most applications.
• Simple setup, operation and maintenance

Accurate Reliable Data Quality
The 3500 series systems significantly improves signal uniformity from instrument-to-instrument, run-to-run, and capillary-to-capillary. With a combination of intelligent hardware, powerful new algorithm and reagent combinations, these methods provide major reduction in the range of signal peak heights obtained across multiple 3500 series instruments (see figure).

Improved Sizing Precision
New thermal sub-system design gives enhanced performance in demanding fragment applications enabling improved sizing precision versus previous generations of capillary electrophoresis platforms.

Ready to Use Consumables With RFID Tags
The 3500 series introduces ready-to-use consumables with pre-packaged polymer pouches, cathode and anode buffer containers, and easy-to-install capillary arrays. Each of these consumables is integrated with RFID tags that enable viewing, tracking, and reporting of critical information about reagents and consumables including usage, lot number, part number, expiry date, and on-instrument lifetime within the 3500 Series Data Collection Software.

Easy to Use Data Collection Software
3500 Series Data Collection software breaks new ground in user-friendly navigation with an intuitive dashboard design, highly visible buttons for common operations, easy-to-read graphical displays to monitor the state of consumables and handy maintenance scheduling calendar functionality. New functionality includes simplified plate set-up and built-in primary analysis with quality control, so you can make decisions about the quality of data as it is produced on the instrument without the need to transfer output files into secondary analysis software packages.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Amperaje15 A
Longitud del capilar50 cm, 36 cm
Requisitos eléctricos100 V to 240 V, 50/60 HZ, 15 A
Ordenador externoHard Drive: 2x 500GB SATA 3.0 Gb/s and 8 MB Data Burst Cache, Memory: 16 GB (2x 8GB) 1600 MHz DDR3 Non-ECC. 4th Gen Intel Core I7 Processor- 3.1 GHz Turbo Processor, Operating System: Windows 7 SP1
Para utilizar con (equipo)POP-6™ Polymer for 3500⁄3500xL Genetic Analyzers, 3500xl Genetic Analyzer Capillary Array (50 cm), POP-4™ Polymer for 3500⁄3500xL Genetic Analyzers, POP-7™ Polymer for 3500⁄3500xL Genetic Analyzers, 3500xl Genetic Analyzer Capillary Array (36 cm)
Formato96-well Plate, 8-tube Strip
Altura exterior (imperial)28.34 in.
Altura exterior (métrico)72 cm
Longitud (imperial) exterior24.01 in.
Longitud (métrico) exterior61 cm
Tensión máx.Electrophoresis Voltage - Up to 20 kV
Número de capilares24 Capillaries
Entorno operativoHumidity: 20%-80% (non-condensing), Room temp: <2° fluctuation during runs, Temperature: 15°C-30°C
Sistema operativoWindows 7 (Service Pack 1)
PolímeroPOP-4™, POP-6™, POP-7™
Consumo de polímerosReplace at earliest of 7 days⁄960 samples⁄50 Injections (960-sample pouches), Replace at earliest of 7 days⁄384 samples⁄20 Injections (384-sample pouches)
Cantidad1 sistema
Longitud de lectura≥850 bp using a 50 cm capillary and POP-7™
Compatibilidad del softwareSequencing Analysis Software, Variant Reporter™ Software, GeneMapper™ Software
Capacidad de procesamientoUp to 1272 samples per day (using run module HID36 POP7)
Garantía1 Year Limited Warranty on Parts and Labor, Service Installation and Application Training Included
Peso82 kg (approximately)
Anchura exterior (imperial)24.01 in.
Anchura exterior (métrico)61 cm
Software TypeData Collection Software, Fragment Analysis
Material de partidaADN
Unit SizeEach
Componentes del envase
4387647
Número de catálogo
4387647
Tamaño de la unidad
Cant.
1
3500xL Genetic Analyzer
Número de catálogo
4405633
Tamaño de la unidad
1 system
Cant.
1
4406933
Número de catálogo
4406933
Tamaño de la unidad
Cant.
1
Contenido y almacenamiento
This package consists of the 24 capillary instrument, the 3500 Data Collection Software, a Dell Workstation and monitor as well as reagent kits for system qualification. This package also includes GeneMapper™ software for fragment analysis, and software for resequencing applications.

Preguntas frecuentes

Is the Plate Septa, 96 well (Cat. No. 4315933) compatible with 3500/3500xL Genetic Analyzers?

Yes, it is compatible with all versions of 3500/3500xL Genetic Analyzers as well as the 3500/3500xL Dx Genetic Analyzers, specifically in RUO mode.

Can I run SNaPshot applications, between 35 and 120 bp, on the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer with a 50 cm capillary array?

Both the SeqStudio Genetic Analyzer and the Applied Biosystems 3500 Genetic Analyzer have run modules specific for the SNaPshot application.

- For the 3500 Genetic Analyzer, the run module is "SNaPshot50_POP7" and it has a ~30 minute run time.
- For the SeqStudio Genetic Analyzer, the run module is "SNaPshot", the optimal size range is 40-120 bp, and it runs for ~25 minutes but the 35 bp fragment will be collected.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

Is there a way to automatically upload my data into the Thermo Fisher Cloud app directly from the Applied Biosystems 3500 Genetic Analyzer, instead of using Data Collection Software?

If you are using the most recent version of Data Collection Software (v3.3), there is a way to automatically upload your data into the Thermo Fisher Cloud app from the instrument. When a run is complete, the data is stored in the results group folder that is specified in the plate record. The data will be automatically uploaded to your account if your User ID is the active ID. Results are saved in your account folders named by the plate and injections, not in the results group folder. Please go to page 43 of the 3500/3500xL Genetic Analyzer User Guide for Data Collection Software v3.3 (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/100079380_3500v3_3RUO_UG.pdf) for details regarding the Thermo Fisher Cloud app.

If you are using Data Collection Software v3.1, you would have to manually upload your data into the Thermo Fisher Cloud app from your data folder on the computer.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.