Applied Biosystems™

SOLiD™ 6-Lane FlowChip

Catalog number:  4461826

 Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SOLiD™ FlowChip is a microfluidic glass device that holds templated beads for ligation sequencing on the 5500⁄5500xl Genetic Analyzers. The SOLiD™ FlowChip Deposition Kit contains reagents for depositing templated beads onto a SOLiD™ FlowChip.

The SOLiD™ FlowChip is specially designed with six precisely cut lanes. Individual samples of templated beads can be deposited into each lane on the FlowChip. Sequencing reagents are delivered to each lane separately, which enables flexible consumption of sequencing reagents for just the number of lanes used for sequencing.

The SOLiD™ FlowChip is notched to set the orientation. Each SOLiD™ FlowChip kit contains one SOLiD™ FlowChip and a deposition cassette that aids in depositing beads.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Product Line: SOLiD™
Product Size: 1 kit
Shipping Condition: Dry Ice
Type: Flow Chip

Contents & storage

Store the SOLiD™ FlowChip at -25°C to -15°C until ready for use.

Documents

Manuals & protocols