5500 SOLiD® Fragment 48 Library Core Kit

Catalog number: 4464415

Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The 5500 SOLiD® Fragment 48 Library Core Kit is used to construct fragment libraries using liquid handling automation for sequencing on 5500 Series genetic analysis systems. This kit contains the necessary reagents (enzymes, buffers, and PCR Amplification mix) for preparing 48 fragment libraries. The kit consists of a 5500 SOLiD® Fragment 48 Library Enzyme Module and a 5500 SOLiD® Fragment 48 Library Amplification Module . Use this kit in conjunction with either the 5500 SOLiD® Fragment Library Standard Adaptors or the appropriate 5500 SOLiD® Fragment Library Barcode Adaptor kit. The 5500 SOLiD™ Fragment 48 Library Core Kit is specifically optimized for use with 5500 Series genetic analysis systems.

Key Benefits:

• Optimized set of reagents for construction of 48 fragment libraries using automation
• Supports amplification-free workflow (depending on desired yield)
• Bead-based cleanup for faster and more efficient processing
• Lower cost per library (compared to SOLiD™ fragment library kits)

Fragment libraries consist of short lengths of DNA specifically selected for optimal performance with a next generation sequencing platform for applications such as targeted or whole genome resequencing. The 5500 SOLiD™ fragment library preparation protocol takes DNA through shearing, end-repair, A-tailing, and ligation with only one intervening bead-based purification that doubles as a size selection step. In many cases, subsequent library amplification is not needed, allowing high complexity libraries to be brought directly into the clonal amplification step.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 5500 Genetic Analyzer, 5500xl Genetic Analyzer
Library Type: Fragment Library
Workflow Step: Library Generation
Starting Material: DNA
Sequencing Type: Genome & DNA Sequencing
Product Line: SOLiD™
Product Size: 48 reactions

Contents & storage

Store at -15°C to -25°C

Documents

Manuals & protocols