5500xl-W Computer Upgrade Kit
Catalog number: 4476413
Applied Biosystems™  Related applications: SOLiD® Next-Generation Sequencing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
This is a computer upgrade kit for the 5500 or 5500xl Genetic Analyzer to optimize performance of the 5500 W upgrade. This kit is a recommended companion to the 5500 to 5500 W and the 5500xl to 5500xl-W upgrade kits that do not include computer upgrade.
This kit includes:
NextIO vCore Express
GPU and RAM upgrades, communication cables
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): 5500 Genetic Analyzer, 5500 W Genetic Analyzer, 5500xl Genetic Analyzer, 5500xl W Genetic Analyzer
Input Voltage: 100-240 VAC
Operating System: Linux
Product Line: SOLiD™
Product Size: 1 each
Type: Computer Accessories
Contents & storage
THIS PRODUCT INCLUDES:
• NextIO vCore Express
• GPU and RAM upgrades, communication cables
Documents
Manuals & protocols