This product is for use with QuantStudio™ 3D Chip Kit v1 (cat. # 4485507 and 4482603 only).
Actual product may vary
Applied Biosystems™
QuantStudio™ 3D Digital PCR UV Sealing Kit
Have Questions?
Catalog number 4488475
Price (USD)/ 1 kit
ERR#1200 - This item cannot be ordered online.
Price (USD)/ 1 kit
ERR#1200 - This item cannot be ordered online.
Add to cart
QuantStudio™ 3D Digital PCR UV Sealing Kit
Catalog number4488475
Price (USD)/ 1 kit
ERR#1200 - This item cannot be ordered online.
-
Add to cart
The QuantStudio™ 3D Digital PCR UV Sealing Kit provides the materials needed to ensure an occlusive seal on the QuantStudio™ 3D Digital PCR Chip following reagent addition. This is accomplished through the application of sealant and the use of UV light to cure the sealant. This kit includes a syringe of sealant and a sealant tip. After loading sample onto the chip, apply the chip lid, add immersion fluid, and seal the fill-hole with the UV sealant and cure the sealant with a UV light pen. If you do do not have a UV pen, an alternate version of this kit is available that includes a UV pen: QuantStudio™ 3D Digital PCR UV Sealing Kit with Pen.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™
Product LineQuantStudio™
Product Size1 kit
TypeUV Sealant Kit
Unit Size1 kit
Contents & Storage
1 x QuantStudio™ 3D UV Cure Chip Sealant 1 x QuantStudio™ 3D UV Cure Chip Sealant Tip Store at room temperature.