Nitenpyram, Thermo Scientific Chemicals
Product Images
Thermo Scientific Chemicals
Nitenpyram, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 463942500
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS150824-47-8
IUPAC Name[(1E)-1-{[(6-chloropyridin-3-yl)methyl](ethyl)amino}-2-nitroethenyl](methyl)amine
Molecular FormulaC11H15ClN4O2
InChI KeyCFRPSFYHXJZSBI-DHZHZOJOSA-N
SMILESCCN(CC1=CC=C(Cl)N=C1)C(\NC)=C\[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Melting point79°C to 85°C
Water=<1 %
Infrared spectrumConforms
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only