Regulatory requirements vary by country; products may not be available in your geographic area.

Cryptosporidium parvum Rapid Test

Catalog number: 5030560

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Cryptosporidium parvum Rapid Test is a lateral immunochromatographic test designed for the detection of Cryptosporidium parvum in young ruminant stool that can also be used with other species. The test is rapid and simple and delivers reliable results. Minimal equipment and hands-on time are required. Results are easy to interpret and available in 10 minutes.
For Veterinary Use Only.

Specifications

Target Organism Class: Cryptosporidium
Detection Method: Colloidal Gold
Disease: Cryptosporidosis
Product Line: Bio-X, Prionics™
Product Size: 10 strips
Species: Cattle⁄Bovine, Goat⁄Caprine, Sheep⁄Ovine

Documents

Manuals & protocols