Regulatory requirements vary by country; products may not be available in your geographic area.

Bovine Immunoglobulins Quantitative ELISA Kit

Catalog number: 5030590

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Bovine Immunoglobulins Quantitative ELISA Kit is a competitive, quantitative ELISA designed for assaying the immunoglobulin concentration in bovine blood serum, milk, and colostrum samples. Assaying the blood serum for immunoglobulins 24 hours after the calf’s first colostrum intake will reveal whether the colostrum has effectively conferred passive immunity on the calves. Diluted test samples and the conjugate— a mixture of bovine immunoglobulins coupled to peroxidase—are simultaneously incubated in 96-well microtiter plates sensitized with protein G specific against bovine immunoglobulins. Binding is detected by a colorimetric reaction after the addition of the substrate.
For Veterinary Use Only.

Specifications

Assay Sensitivity: See product documentation
Gene ID: Non-Gene
Gene Symbol: Non-Gene
Immunoassay Kit Format: Various Other
Product Line: Bio-X, Prionics™
Product Size: 80 samples
Target (Kit Named): Bovine Immunoglobulins
Target Species (Named): Bovine
Target Species (Validated): Bovine

Documents

Manuals & protocols