Regulatory requirements vary by country; products may not be available in your geographic area.

Epsilon Toxin Rapid Test Kit

Catalog number: 5030610

 Related applications: Animal Health

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Epsilon Toxin Rapid Test Kit is a lateral immunochromatographic test designed to detect Clostridium perfringens Epsilon Toxin in biological fluids or in culture supernatants. The use of monoclonal antibody as conjugate to detect Epsilon toxin enables excellent specificity and very reliable results. The test requires minimal hands-on-time and results are typically available in 10 minutes.
For Veterinary Use Only.

Specifications

Target Organism Class: C. perfringens
Detection Method: Colloidal Gold
Disease: Enterotoxaemia
Product Line: Bio-X, Prionics™
Product Size: 10 strips
Species: All Species

Documents

Manuals & protocols