Thermo Scientific™
UltiMate™ HPG-3400SD Standard Binary Pump
 Related applications:
Industrial Chromatography
Increase your binary method flexibility. The Thermo Scientific™ Dionex™ UltiMate™ HPG-3400SD Standard Binary Pump allows four different mobile phases, two at a time. Thermo Scientific™ SmartFlow™ technology delivers pulse-free flows without a damper to ensure precise retention times. With support for pressures up to 62MPa (9,000 psi/620 bar) at flow rates up to 10mL/min., UltiMate 3000 SD pumps excel in standard LC applications and offer full compatibility with fast UHPLC separations, including short sub-2 µm particle columns for faster analysis and lower solvent consumption.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Recommended Use
This pump is recommended for use within the UltiMate 3000 Binary Standard system. The HPG-3400SD pump is ideal for routine analyses with an added measure of flexibility using conventional LC and high-throughput LC in the pharmaceutical, food and beverage and environmental industries.
Simple Method Transfer
UltiMate 3000 Standard SD and Rapid Separation (RS) pumps and system configurations provide similar gradient delay and extra-column volumes. This simplifies the method transfer between method development labs commonly using UHPLC pumps and QC and production labs with their standard LC pump setups.
Features
  • Serial dual-piston operating principle, high-pressure gradient proportioning
  • Wide operating flow rate range from 100µL to 10mL/min. with high flow rate accuracy
  • Support of operating pressures of up to 62 MPa (9,000 psi/620 bar)
  • Thermo Scientific™ SmartFlow™ technology to automatically compensate for compressibility;no manual solvent setting needed
  • Large selection of mixing volumes to perfectly balance gradient delay volume with mixing efficiency
  • Active rear-seal wash and floating pistons to maximize seal lifetime and pump uptime
Documents
Manuals & protocols