Interested in a bulk or custom version of this product? Request pricing.

EREG Recombinant Mouse Protein

Catalog number: 50599M01H250

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Epiregulin (EREG), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse EREG (Q61521) (Val 56-Leu 101).

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Angiogenesis, Cell Cycle & Proliferation
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: EPR, MGC36144
Gene Symbol: EREG
Molecular Weight: 33.8 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Family: Growth Factors & Receptors
Protein Name: Proepiregulin
Protein Subtype: EGF (Epidermal Growth Factor), Growth Factor Receptors
Protein Tag or Fusion: Untagged
Purity: ≥ 90% by SDS-PAGE
Species: Mouse
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols

Recommended products