Search Thermo Fisher Scientific
Catalog Number | Description | For Use With (Application) | For Use With (Equipment) | Type |
---|---|---|---|---|
6036.2305 | Viper Kit for Vanquish Duo for Dual LC | Two of the same of two different assays simultaneously | Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems | Advanced Capillary Kit |
6036.0100 | Extension Kit for Automated Method Scouting for Vanquish LC Systems | Method Development | Vanquish Core HPLC System, Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems | Extension Kit |
6036.2030 | Viper Kit for Trap Heart-Cut and Simple Switch 2D-LC for Vanquish UHPLC Systems | 2D-LC | Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems | Advanced Capillary Kit |
6036.2020 | Viper Kit for Vanquish Duo for Tandem LC and LC-MS | Intelligent column switching for maximized detector usage | Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems | Advanced Capillary Kit |
6036.2010 | Viper Kit for Vanquish Duo for Inverse Gradient | Standard- free quantitation with charged aerosol detection | Vanquish Flex UHPLC Systems, Vanquish Horizon UHPLC Systems | Advanced Capillary Kit |
6036.2807 | Viper Kit for Automated Method Scouting for Vanquish UHPLC Systems | Method Development | Vanquish Flex and Horizon UHPLC Systems | Advanced Capillary Kit |
Improve chromatographic resolution by minimizing dead volume with the Thermo Scientific™ Viper™ Fingertight Fitting Systems. These pre-assembled kits allow you to quickly connect LC modules, valves, and columns for leak-free fluidic HPLC and UHPLC connections. Ideally suited for running application-specific workflows with your HPLC and LC-MS systems, like the Thermo Scientific™ Vanquish™ Method Development LC Systems, Thermo Scientific Vanquish Online 2D-LC Systems, or Thermo Scientific Vanquish Duo UHPLC Systems.