Search Thermo Fisher Scientific
Ensure virtually zero-dead-volume connections for optimal nano LC performance with easy-to-use Thermo Scientific™ nanoViper™ Fingertight Fittings.
Catalog Number | Description |
---|---|
6041.5575 | nanoViper 50 μm x 650 mm |
6041.5580 | nanoViper 50 μm x 750 mm |
6041.5560 | nanoViper 50 μm x 550 mm |
6041.5540 | nanoViper 50 μm x 350 mm |
6041.5125 | nanoViper 50 μm x 950 mm |
6041.5124 | nanoViper 50 μm x 150 mm |
6041.5123 | nanoViper 50 μm x 70 mm |
Maximize nano liquid chromatography (LC) performance and convenience with Thermo Scientific™ nanoViper™ Fingertight Fittings. Virtually zero-dead-volume connections ensure optimal separation efficiencies. Preassembled and finger-tightened, nanoViper fingertight fittings enable easy, tool-free instrument setup. These fittings are high-pressure tested to provide UHPLC compatibility up to 1200 bar (17,400 psi).
Robust LC Performance
Easy to Install
Flexible
Recommended Use
Thermo Scientific™ UltiMate™ 3000 RSLCnano Systems are equipped with the nanoViper Fingertight Fitting System as a standard.