Search Thermo Fisher Scientific
Catalog Number | For Use With (Equipment) | Type | Material |
---|---|---|---|
6074.0280 | UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS) | Capillary Flow Cell | Fused Silica |
6074.0320 | Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS) | Semi-Preparative Flow Cell | PEEK |
6074.0200 | UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS) | Analytical Flow Cell | PEEK |
6074.0250 | UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS) | Analytical Flow Cell | Stainless Steel |
6074.0270 | UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS) | Nano Flow Cell | Fused Silica |
6074.0285 | Dionex UltiMate 3000 Variable Wavelength Detector | Ultra-Low Dispersion UV-Monitor | Fused Silica |
6074.0290 | Dionex UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS) | Mic Flow Cell | Fused Silica |
6074.0300 | UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS) | Semi-Micro Flow Cell | PEEK |
6074.0360 also known as 6074.036 | UltiMate 3000 Variable Wavelength Detector, VWD-3x00(RS) | Semi-Micro Flow Cell | Stainless Steel |