Dynabeads™ mRNA DIRECT™ Purification Kit
Invitrogen™

Dynabeads™ mRNA DIRECT™ Purification Kit

The Dynabeads™ mRNA DIRECT™ Kit is designed for simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
6101210 mL
610115 mL
Catalog number 61012
Price (USD)
1,394.65
Online exclusive
1,516.00
Save 121.35
Each
Quantity:
10 mL
Price (USD)
1,394.65
Online exclusive
1,516.00
Save 121.35
Each

The Dynabeads™ mRNA DIRECT™ Kit is designed for simple and rapid isolation of pure, intact polyadenylated (polyA) mRNA directly from the crude lysate of animal and plant cells and tissues. The isolated mRNA is suitable for use in all downstream applications.

Advantages of the Dynabeads mRNA DIRECT Kit:

  • Fast – 15 minute procedure yields pure, intact mRNA
  • Highly pure mRNA isolation – best choice upstream of cDNA synthesis
  • Sensitive mRNA isolation – enables cDNA synthesis and cDNA library construction from ultra-small starting samples (enables cDNA library construction from a single cell)

System overview
The isolation protocol relies on base pairing between the polyA residues at the 3' end of most mRNA, and the oligo (dT)25 residues covalently coupled to the surface of the Dynabeads. Other RNA species lacking a polyA tail will not hybridize to the beads and are readily washed away. Ribosomal RNA, DNA, proteins, and small RNA molecules (such as transfer RNA, micro RNA, and small nucleolar RNA) do not bind to the beads and are discarded. RNase inhibiting agents in the Lysis/Binding Buffer together with stringent hybridization and washing conditions ensure the isolation of pure, intact mRNA from crude samples rich in RNase, without the use of strong chaotropic agents. The mRNA purification beads specifically target and capture the mRNA transcriptome from an extremely wide variety of crude starting samples (see protocol). 1 mg of Dynabeads Oligo (dT)25 beads (200 μL) binds up to 2 μg of mRNA. A typical mammalian cell contains about 10–30 pg of total RNA, from which 1–5% is mRNA.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Elution Volume10 to 100 μL
Final Product TypemRNA
For Use With (Application)RT-PCR, qPCR, cDNA library construction, microarray analysis
For Use With (Equipment)Automated Liquid Handling Systems
High-throughput CompatibilityHigh-throughput Compatible
Quantity10 mL
Shipping ConditionRoom Temperature
Starting Material AmountCells: ≤106
Plant: ≤400 mg
Tissue: ≤200 mg
Yield2 μg mRNA per 200 μL of beads (Binding capacity)
Isolation TechnologyMagnetic Bead
Sample TypeCells, Plants, Plasma, Serum, Tissue, Total RNA, Blood
Unit SizeEach
Contents & Storage

• 10 mL Dynabeads Oligo (dt)25, 4°C
• 60 mL Lysis/Binding Buffer; 4°C
• 120 mL Washing Buffer A; 4°C
• 60 mL Washing Buffer B; 4°C
• 15 mL 10 mM Tris-HCl; 4°C

Frequently asked questions (FAQs)

I am getting DNA contamination after mRNA isolation using Dynabeads magnetic beads. Why is this?

There are several reasons why DNA contamination may occur:

- Incomplete DNA shearing.
- Incomplete removal of sample lysate after the hybridization step.
- Insufficient washing and/or removal of wash buffers.
- The ratio of sample to beads was too high.

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Citations & References (6)

Citations & References
Abstract
Extracellular plasma RNA from colon cancer patients is confined in a vesicle-like structure and is mRNA-enriched.
Authors:García JM, García V, Peña C, Domínguez G, Silva J, Diaz R, Espinosa P, Citores MJ, Collado M, Bonilla F
Journal:RNA
PubMed ID:18456845
'Little is yet known about the origin and protective mechanism of free nucleic acids in plasma. We investigated the possibility of these free nucleic acids being particle associated. Plasma samples from colon cancer patients and cell culture media were subjected to various antibody incubations, ultracentrifugation, and RNA extraction protocols for ... More
Relationships and differentially expressed genes among pancreatic cancers examined by large-scale serial analysis of gene expression.
Authors:Ryu B, Jones J, Blades NJ, Parmigiani G, Hollingsworth MA, Hruban RH, Kern SE
Journal:Cancer Res
PubMed ID:11830538
'Pancreatic adenocarcinoma is among the most fatal of cancers, in part because of late diagnosis and a lack of effective therapies. Comprehensive studies are needed to better understand and address the cellular mechanisms and pathways of tumorigenesis. Serial analysis of gene expression was used to analyze gene expression profiles of ... More
Stem cell and epithelial-mesenchymal transition markers are frequently overexpressed in circulating tumor cells of metastatic breast cancer patients.
Authors:Aktas B, Tewes M, Fehm T, Hauch S, Kimmig R, Kasimir-Bauer S
Journal:Breast Cancer Res
PubMed ID:19589136
The persistence of circulating tumor cells (CTC) in breast cancer patients might be associated with stem cell like tumor cells which have been suggested to be the active source of metastatic spread in primary tumors. Furthermore, these cells also may undergo phenotypic changes, known as epithelial-mesenchymal transition (EMT), which allows ... More
The transcription factors SOX9 and SOX10 are vitiligo autoantigens in autoimmune polyendocrine syndrome type I.
Authors:Hedstrand H, Ekwall O, Olsson MJ, Landgren E, Kemp EH, Weetman AP, Perheentupa J, Husebye E, Gustafsson J, Betterle C, Kämpe O, Rorsman F
Journal:J Biol Chem
PubMed ID:11423552
Vitiligo is common in the hereditary disorder autoimmune polyendocrine syndrome type I (APS I). Patients with APS I are known to have high titer autoantibodies directed against various tissue-specific antigens. Using sera from APS I patients for immunoscreening of a cDNA library from human scalp, we identified the transcription factors ... More
Disease-associated mutations in human mannose-binding lectin compromise oligomerization and activity of the final protein.
Authors:Larsen F, Madsen HO, Sim RB, Koch C, Garred P
Journal:J Biol Chem
PubMed ID:14764589
Deficiency of human mannose-binding lectin (MBL) caused by mutations in the coding part of the MBL2 gene is associated with increased risk and severity of infections and autoimmunity. To study the biological consequences of MBL mutations, we expressed wild type MBL and mutated MBL in Chinese hamster ovary cells. The ... More