Red Filter for myECL™ Imager
Catalog number: 62240
Thermo Scientific™  Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Thermo Scientific 600-640nm bandpass filter is a threaded, 50mm-diameter glass filter. This product is used for the addition of a red filter to one of two open positions in the myECL Imager for detection (emission) wavelengths of common fluorescent protein stains, such as SYPRO* Red Protein Gel Stain, SYPRO Ruby Protein Gel Stain and Deep Purple* Total Protein Stain.
More Product Data
Improved Western blot detection using CCD digital imaging
Related Products
myECL™ Imager
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): myECL™ Imager
Product Line: myECL™
Product Size: 1 filter
Type: Filter
Contents & storage
0.0
Documents
Manuals & protocols