UV Exposure Screen for myECL™ Imager

Catalog number: 62244

Thermo Scientific™  Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This clear plastic sample screen is used to cover the transilluminator on the Thermo Scientific myECL Imager. It is a replacement for the accessory used on the transilluminator for acquiring images in the UV mode. Measurements: 24.5 cm x 19.5 cm x 3.2mm.

More Product Data
Improved Western blot detection using CCD digital imaging

Related Products
myECL™ Imager
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): myECL™ Imager
Product Line: myECL™
Product Size: 1 screen
Type: Other Accessories

Contents & storage

0.0

Documents

Manuals & protocols