CaptureSelect™ Biotin Anti-IgG3 (Human) Conjugate
Thermo Scientific™
CaptureSelect™ Biotin Anti-IgG3 (Human) Conjugate
Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG3 (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG3 subclass, most likely the hinge region.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7103042100100 μg
7103042500500 μg
Catalog number 7103042100
Price (USD)
636.00
Each
Quantity:
100 μg
Price (USD)
636.00
Each
CaptureSelect Biotin Anti-IgG3 (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG3 subclass, most likely the hinge region. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Biotin Anti-IgG3 (Hu) Conjugate allows you to:
  • Detect, quantitate, and characterize human IgG3 antibodies
  • Avoid cross-binding with other human IgG subclasses like IgG1, 2 and 4 and isotypes like IgM and IgA including IgG from bovine sources such as FCS
Applications for CaptureSelect Biotin Anti-IgG3 (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateBiotin and Other Haptens
Product LineCaptureSelect
Product TypeBiotin Anti-IgG3 Conjugate
Quantity100 μg
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).