Interested in a bulk or custom version of this product? Request pricing.

MMP9 Recombinant Rat Protein, His Tag

Catalog number: 80049R08H5

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Matrix Metallopeptidase 9 (MMP9), recombinant rat protein is supplied in frozen format. It is suitable for use in protein studies such as enzymetic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the rat MMP9 (EDL96479.1) (Met 1-Pro 708) fused to a polyhistidine tag at the C-terminus.

Activity: Measured by its ability to cleave a fluorogenic peptide substrate Mca-KPLGL-Dpa-AR-NH2 (R&D Systems, Catalog # ES010). The specific activity is >1000 pmoles/min/µg.

Formulation: 50 mM MES, 100 mM NaCl, 1 mM CaCl2, 0.2 µg/µl BSA, 10% glycerol, pH7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Angiogenesis, Cardiovascular Research, Drug Development, Inflammation
Endotoxin Level: < 1 EU/µg
Expression: Mammalian
Form: Frozen
Gene Aliases: Mmp9
Gene Symbol: MMP9
Molecular Weight: 77.8 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Name: matrix metallopeptidase 9
Protein Subtype: Proteases
Protein Tag or Fusion: His Tag
Purity: ≥ 90% by SDS-PAGE
Species: Rat
Validated Application: Assay Standard

Contents & storage

Store at -80 °C.

Documents

Manuals & protocols