CaptureSelect™ Ig Select Leakage ELISA Kit

Catalog number: 810083510

 Related applications: Antibody Purification | Contaminant & Impurity QC Testing | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CaptureSelect™ IgSelect Leakage ELISA detects IgSelect affinity ligand contamination of human IgG purified using IgSelect affinity resin (GE Healthcare). Even when covalently attached, small amounts of the IgSelect affinity ligand can leach off of the chromatography support and co-elute with the antibody. This ELISA provides sensitivity to detect IgSelect affinity ligand contamination of less than 1 ng/mL. The assay is designed to minimize interference and to provide accurate quantitation in the presence of human IgG. The CaptureSelect™ IgSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: ELISA
Target: IgSelect Affinity Ligand
For Use With (Equipment): Microplate Reader
Product Line: CaptureSelect™
Product Size: 10 assays

Contents & storage

Store at -5 to -30°C.

Documents

Manuals & protocols