Nicolet™ Condensed Phase Academic Sampler Library
Thermo Scientific™

Nicolet™ Condensed Phase Academic Sampler Library

Suited to academic and small QC labs, this library includes spectra illustrating the major functional groups and combinations of commonly observed functional groups.
Have Questions?
Catalog NumberType
834-009800Nicolet Condensed Phase Academic Sampler Library
Catalog number 834-009800
Price (USD)
-
Type:
Nicolet Condensed Phase Academic Sampler Library

The Thermo Scientific™ Nicolet Condensed Phase Academic Sampler is suited to the needs of academic institutions and small QC labs. It includes spectra that illustrate the major functional groups and combinations of commonly observed functional groups.

Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.

Chosen by chemistry professors from many disciplines, this high resolution format library includes spectra of chemicals used in a wide range of common laboratory experiments.
  • Spectra illustrate the major functional groups and combinations of functional groups commonly observed in academic chemistry laboratories
  • Included chemicals are also important building blocks commonly found in industrial applications
  • Contains 1,000 spectra
  • Data format: High Resolution (HR)
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeNicolet Condensed Phase Academic Sampler Library
Unit SizeEach