Thermo Scientific™
Aldrich™ Esters, Lactone, Anhydrides Spectral Library
 Related applications:
Lab Automation
Efficiently identify compounds and bring greater effeciencies to your lab with the The Thermo Scientific™ Aldrich™ Esters, Lactone, Anhydrides Spectral Library, a subset of the Aldrich FT-IR Spectral Library.
Take the guesswork out of your spectral analysis. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
This Aldrich Material-Specific FT-IR Library collection represents a wide variety of most common chemicals from The Aldrich Handbook of Fine Chemicals, divided by similar functional groups. These spectra were assembled from the Aldrich Collections of FT-IR Spectra Editions I or II, and the data has been carefully examined and processed by Thermo Fisher Scientific.
  • Contains 1,653 spectra
  • Data format: High Resolution (HR)
About Spectral Libraries:
  • Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
  • Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification&emdash;or at least classification&emdash;of the material.
  • Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Documents
Manuals & protocols