Product Image
Invitrogen™
MagniSort™ Mouse CD90.1 Positive Selection Kit
The MagniSort™ Mouse CD90.1 Positive Selection Kit is designed for the magnetic separation of CD90.1+ cells by positive selection. ItRead more
Catalog NumberQuantity
8802-6843-74200 tests
Catalog number 8802-6843-74
Price (JPY)
101,400
Online offer
Ends: 27-Mar-2026
169,100
Save 67,700 (40%)
Each
Quantity:
200 tests
The MagniSort™ Mouse CD90.1 Positive Selection Kit is designed for the magnetic separation of CD90.1+ cells by positive selection. It has been optimized for the isolation of CD90.1+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD90.1 antibody and streptavidin-coated magnetic beads. CD90.1+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD90.1+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse/Rat CD90.1, clone HIS51.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD90.1+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach