MagniSort™ Mouse CD11c Positive Selection Kit
Invitrogen™
MagniSort™ Mouse CD11c Positive Selection Kit
The MagniSort™ Mouse CD11c Positive Selection Kit is designed for the magnetic separation of CD11c+ cells by positive selection. ItRead more
Have Questions?
Catalog NumberQuantity
8802-6861-74200 tests
Catalog number 8802-6861-74
Price (KRW)
912,000
Online offer
Ends: 31-Dec-2026
1,072,000
Save 160,000 (15%)
Each
Quantity:
200 tests
Price (KRW)
912,000
Online offer
Ends: 31-Dec-2026
1,072,000
Save 160,000 (15%)
Each
The MagniSort™ Mouse CD11c Positive Selection Kit is designed for the magnetic separation of CD11c+ cells by positive selection. It has been optimized for the isolation of CD11c+ cells from mouse spleens utilizing a biotinylated Anti-Mouse CD11c antibody and streptavidin-coated magnetic beads. CD11c+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD11c+ cells by decanting.
After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD11c, clone N418 and Anti-Mouse CD11b, clone M1/70.
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeCD11c+
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypePositive Selection Kit
Unit SizeEach