ActivX™ TAMRA-FP Serine Hydrolase Probe
ActivX™ TAMRA-FP Serine Hydrolase Probe
ActivX™ TAMRA-FP Serine Hydrolase Probe
Thermo Scientific™

ActivX™ TAMRA-FP Serine Hydrolase Probe

ActivX™ probes have been designed to selectively label and enrich serine hydrolases through binding of their respective active sites with tagged fluorophosphonate to enable assessment of enzyme activity.
Have Questions?
Catalog NumberQuantity
883186.8 μg
Catalog number 88318
Price (USD)
606.00
Each
Quantity:
6.8 μg
Price (USD)
606.00
Each

The TAMRA-FP probe feature a reactive fluorophosphonate group that is used to label and detect active serine hydrolases by fluorescent gel imaging, capillary electrophoresis or mass spectrometry. An anti-TAMRA antibody is also available for immuno-enrichment of TAMRA-FP probe-labeled proteins.

ActivX™ active-site enrichment probes are advantageous for determining active enzyme levels through the use of analog-tagged probes to allow profiling of both inactive and active enzymes. The ActivX probes selectively enrich only those enzymes that are functionally active and biologically relevant at the time of labeling. With selective enrichment, one can identify and profile target enzyme classes across samples or can assess the specificity and affinity of enzyme inhibitors.

The serine hydrolase superfamily is one of the largest, most diverse enzyme families in eukaryotic proteomes. Serine hydrolases are generally grouped into two large families: serine proteases (e.g., trypsin, elastase and thrombin) and metabolic serine hydrolases. Metabolic serine hydrolases are divided into multiple enzyme subclasses (e.g., esterases, lipases, amidases and peptidases) based on structure, catalytic mechanism and substrate preference. These probes can be used to assess activity or screen small molecule inhibitors against enzymes derived from cell lysates, subcellular fractions, tissues and recombinant proteins.

These products are subject to a limited use label license.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
AssayProtein Quantitation Assay
DescriptionActivX™ TAMRA-FP Serine Hydrolase Probe
Detection MethodChemiluminescence, Mass Spectrometry, Western Blot
For Use With (Application)Pull-Down
FormatTube, Spin Column
FormulationTAMRA-fluorophosphonate, MW 680
Label or DyeOther Label(s) or Dye(s)
Quantity6.8 μg
Substrate TypeSerine Hydrolase Substrate
TargetSerine Hydrolase
Product LineActivX
TypeFluorophosphonate (FP) probe
Unit SizeEach
Contents & Storage
Store at -80°C away from light. Stock solutions in DMSO can be stored at -20°C for 6 months.

Frequently asked questions (FAQs)

Can I isolate my proteins based on their post-translational modification (PTM) or active binding site?

Enrichment of specific proteins or protein complexes can most easily be accomplished by using target-specific immobilized antibody or PTM specific affinity binding. We offer several kits targeting phosphorylated proteins, alpha- linked mannose and terminal glucose residues, N-acetyl glucosamine, polyubiquinated proteins, ATP binding sites, and active serine hydrolase enzymes. Choose the right protein enrichment kit for your application using the following link (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/protein-enrichment.html).

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.