Interested in a bulk or custom version of this product? Request pricing.

TP53 (P53) Recombinant Cynomolgus Protein

Catalog number: 90001CNAE25

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Tumor protein p53 (P53), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the cynomolgus (Macaca fascicularis) P53 (E3U906) (Met 1-Asp 393).

Activity:

1. Measured by its ability to bind biotinylated human S100B in a functional ELISA.

2. Measured by its ability to bind biotinylated human RRM2B in a functional ELISA.

Formulation: Lyophilized in 100 mM Tris, 3% Glycerol, 4 mM DTT, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Lyophilized
Gene Aliases: p53, TP53
Gene Symbol: TP53
Molecular Weight: 43.7 kDa
Product Line: Sino Biological®
Product Size: 5 x 5 µg
Protein Family: Other Proteins
Protein Name: Cynomolgus Cellular Tumor Antigen p53
Protein Subtype: Other Proteins
Protein Tag or Fusion: Untagged
Purity: ≥ 85% by SDS-PAGE
Research Category: Apoptosis, Cell Cycle & Proliferation, Oncology, Signal Transduction
Species: Monkey
Validated Application: Assay Standard

Contents & storage

Includes 5 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols