Axiom™ Exome 319 Array Plate
Applied Biosystems™

Axiom™ Exome 319 Array Plate

Streamline your causal variant studies by enriching genome-wide association studies (GWAS) datasets with an extensive set of novel, rare, putativeRead more
Have Questions?
Catalog NumberQuantity
9021061 plate
Catalog number 902106
Price (USD)
-
Quantity:
1 plate
Streamline your causal variant studies by enriching genome-wide association studies (GWAS) datasets with an extensive set of novel, rare, putative functional variants. The content was discovered in 16 major exome sequencing projects spanning a total of 12,000 samples.

With the Axiom Exome 319 Array Plate, you get:
• Comprehensive content for highest discovery power
   - More than 300,000 coding SNPs including non-synonymous and synonymous SNPs as well as variants in splice and stop codons.
   - Approximately 30,000 single-base and complex indels from the draft Phase 1 1000 Genomes Project exome calls.
• Informative markers to enable integrated data analysis
   - AIMs: Ancestry informative markers to measure Latino and African American ancestry. All markers have low linkage disequilibrium (LD) and span all autosomes and chromosome X.
   - >GWAS tags: Includes over 5,000 SNPs reported in the NHGRI Catalog of Published Associations (August 2011) and additional unpublished hits from scientific collaborations.
   - High value: Contains other markers of interest, including DNA fingerprinting SNPs, human leukocyte antigen (HLA) genes, chromosome Y, mitochondrial DNA, and microRNA sequences.
• Customization to include your candidate genes
   - Include an additional 100,000 SNPs from your own candidate genes to meet your research needs. Choose from the Axiom Genomic Database of 11M genotype-tested markers, or include variants of your own choosing.
• Proven performance
   - The panel has been fully genotype-tested against stringent QC criteria to ensure high-quality data.
   - Every marker has been tested against approximately 1,200 samples to ensure high sample pass, call rate, and reproducibility.

Axiom Exome Genotyping Array Plates include:
   - Axiom 2.0 Reagent Kit for automated or manual processing of up to 96 samples per array plate.
   - A fully automated workflow with validated robotics methods for target preparation and array processing, based on the Beckman Biomek™ FXP Target Prep Express System and the GeneTitan™ MC Instrument.
   - A manual target preparation protocol for lower-throughput users.
   - Axiom Analysis Suite for automated allele calling and easier quality assessment of called genotypes.

Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesHuman
Unit SizeEach

Frequently asked questions (FAQs)

Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?

The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.