QConnect Serology Multimarker Controls
Thermo Scientific™  Related applications: Diagnostic Testing
Estimate the precision and reproducibility of serological assays that test human plasma from donations of whole blood and blood components with the Thermo Scientific™ QConnect Serology Multimarker Controls. These controls mimic naturally occurring plasma specimens with low reactivity of multiple analytes and enable monitoring of day-to-day test performance of various combinations of IVD assays for the detection of multiple human infectious diseases. The controls are for In Vitro Diagnostic Use.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
Showing {{showMoreLimit}} of {{commerceData.products.length}} |  Showing {{shownProducts.length}} of {{commerceData.products.length}} | 
Back to top
Description
Estimate the precision and reproducibility of serological assays that test human plasma from donations of whole blood and blood components with the Thermo Scientific™ QConnect Serology Multimarker Controls. These controls mimic naturally occurring plasma specimens with low reactivity of multiple analytes and enable monitoring of day-to-day test performance of various combinations of IVD assays for the detection of multiple human infectious diseases. The controls are for In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.
Documents
Manuals & protocols