ACMA (9-amino-6-chloro-2-methoxyacridine) is a DNA intercalator that selective binds to poly (d(A-T)). Excitation of the ACMA-DNA complex (excitation/emission maxima ∼419/483Read more
Have Questions?
Catalog Number
Quantity
A1324
100 mg
Catalog number A1324
Price (USD)
240.26
Special offer
Online exclusive
Ends: 31-Dec-2026
293.00
Save 52.74
Each
Quantity:
100 mg
Price (USD)
240.26
Special offer
Online exclusive
Ends: 31-Dec-2026
293.00
Save 52.74
Each
ACMA (9-amino-6-chloro-2-methoxyacridine) is a DNA intercalator that selective binds to poly (d(A-T)). Excitation of the ACMA-DNA complex (excitation/emission maxima ∼419/483 nm) is possible with most UV-light sources, making it compatible for use with both shorter- and longer-wavelength dyes. ACMA also apparently binds to membranes in the energized state and becomes quenched if a pH gradient forms. It has been extensively employed to follow cation and anion movement across membranes and to study the proton-pumping activity of various membrane-bound ATPases.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorBlue
Detection MethodFluorescence
Excitation Wavelength Range419⁄483
For Use With (Equipment)Fluorescence Microscope
Quantity100 mg
Shipping ConditionRoom Temperature
Label TypeFluorescent Dye
Product TypeNucleic Acid Stain
Sub Cellular LocalizationCell Membranes, Lipids, Nucleus
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.
Citations & References (84)
Citations & References
Abstract
Saccharomyces cerevisiae lacking Btn1p modulate vacuolar ATPase activity to regulate pH imbalance in the vacuole.
Authors:Padilla-López S, Pearce DA
Journal:J Biol Chem
PubMed ID:16423829
'The vacuolar H(+)-ATPase (V-ATPase) along with ion channels and transporters maintains vacuolar pH. V-ATPase ATP hydrolysis is coupled with proton transport and establishes an electrochemical gradient between the cytosol and vacuolar lumen for coupled transport of metabolites. Btn1p, the yeast homolog to human CLN3 that is defective in Batten disease, ... More
Yeast V-ATPase complexes containing different isoforms of the 100-kDa a-subunit differ in coupling efficiency and in vivo dissociation.
Authors:Kawasaki-Nishi S, Nishi T, Forgac M
Journal:J Biol Chem
PubMed ID:11278748
'The 100 kDa a-subunit of the yeast vacuolar (H(+))-ATPase (V-ATPase) is encoded by two genes, VPH1 and STV1. These genes encode unique isoforms of the a-subunit that have previously been shown to reside in different intracellular compartments in yeast. Vph1p localizes to the central vacuole, whereas Stv1p is present in ... More
Subunit D (Vma8p) of the yeast vacuolar H+-ATPase plays a role in coupling of proton transport and ATP hydrolysis.
Authors:Xu T, Forgac M
Journal:J Biol Chem
PubMed ID:10801866
'To investigate the function of subunit D in the vacuolar H(+)-ATPase (V-ATPase) complex, random and site-directed mutagenesis was performed on the VMA8 gene encoding subunit D in yeast. Mutants were selected for the inability to grow at pH 7.5 but the ability to grow at pH 5.5. Mutations leading to ... More
Oligodeoxynucleotides covalently linked to intercalating agents: a new family of gene regulatory substances.
Authors:Héléne C, Montenay-Garestier T, Saison-Behmoaras T, Toulmé JJ, Boidot-Forget M, Cazenave C, Asseline U, Lancelot G, Maurizot JC, Toulmé F
Journal:Biochem Soc Trans
PubMed ID:3709940
Interactions between the F1 and F0 parts in the Escherichia coli ATP synthase. Associations involving the loop region of C subunits.
Authors:Watts SD, Capaldi RA
Journal:J Biol Chem
PubMed ID:9182524
'The N-ethylmaleimide reactivity of c subunits in Escherichia coli F1F0 ATP synthase (ECF1F0) isolated from five mutants, each with a cysteine at a different position in the polar loop region (positions 39, 40, 42, 43, and 44), has been investigated. The maleimide was found to react with Cys placed at ... More