4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N?-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N′-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Andrew W. Nicholls; Kazuki Akira; John C. Lindon; R. Duncan Farrant; Ian D. Wilson; John Harding; David A. Killick; and Jeremy K. Nicholson. NMR Spectroscopic and Theoretical Chemistry Studies on the Internal Acyl Migration Reactions of the 1-O-Acyl-β-d-glucopyranuronate Conjugates of 2-, 3-, and 4-(Trifluoromethyl)benzoic Acids. Chem. Res. Toxicol. 1996, 9 (8), 1414-1424.
- C. U. Galdiga; Tyge Greibrokk. Ultra trace determination of fluorinated aromatic carboxylic acids in aqueous reservoir fluids by solid phase extraction in combination with negative ion chemical ionisation mass spectrometry after derivatisation with pentafluorobenzyl bromide. Fresenius' Journal of Analytical Chemistry. 1998, 361 (8), 797-802.