Oncomine™ Immune Response Research Assay
Ion Torrent™

Oncomine™ Immune Response Research Assay

The Oncomine™ Immune Response Research Assay is a targeted gene expression assay designed for the Ion™ next-generation sequencing (NGS) platform.Read more
Have Questions?
Catalog NumberQuantity
A3288132 Samples
Catalog number A32881
Price (USD)
12,690.00
Each
Quantity:
32 Samples
Price (USD)
12,690.00
Each

The Oncomine™ Immune Response Research Assay is a targeted gene expression assay designed for the Ion™ next-generation sequencing (NGS) platform. This pan-cancer gene expression assay is designed to interrogate the tumor microenvironment to enable mechanistic studies and identification of predictive biomarkers for immunotherapy in retrospective or prospective clinical trial research cohorts. The assay is optimized to measure the expression of genes involved in tumor-immune interactions, including the low-expressing genes involved in inflammatory signaling.

This assay contains the reagents for manual library construction and a single pool of primers used to perform multiplex PCR for preparation of amplicon libraries from formalin-fixed paraffin-embedded (FFPE) samples. The assay enables quantitative evaluation of the expression of markers associated with different leukocyte subsets, antigen presentation, checkpoint pathways, and tumor progression. This 400-gene assay is supported with a user-friendly informatics workflow and demonstrates high sensitivity for low-expressing transcripts derived from FFPE samples (request a gene list). The Oncomine Immune Response Research Assay leverages Ion AmpliSeq™ technology to deliver results from as little as 10 ng of total RNA, enabling robust performance from challenging FFPE samples.

Assay workflow

The Oncomine Immune Response Research Assay manual workflow features high sample multiplexing on Ion S5™ and Ion PGM™ sequencing systems. The entire workflow, from sample extraction to results, can be completed in less than 48 hours. Note: An alternate assay is available (Cat. No. A32928) for the preparation of Ion AmpliSeq libraries in automated mode through use of an Ion Chef™ System.

Assay performance

  • High repeatability and sensitive detection of gene expression across different types of solid tumors
  • Low sample input requirement (10 ng of total RNA material)
  • High sensitivity detection of low-expressing signaling molecules such as interleukins and interferon gamma
  • High sample multiplexing with flexibility to run 8 samples per single run
  • Efficient workflow that offers sample-to-results in less than 48 hours
  • Automated and seamlessly integrated analysis solution
  • Interpretation of immune surveillance and tumor progression mechanism with functional annotation of genes and pathways

Informatics

The Oncomine Immune Response Research Assay comes with optimized informatics and visualization software that minimizes the complexity of data analysis. The analysis workflow consists of a data exploration component automated by the Immune Response Torrent Suite™ Plug-in and a two-group differential analysis provided by the Affymetrix™ Transcriptome Analysis Console (TAC). Together these tools offer a streamlined process for common gene expression analyses and exportable data files that are amenable to further exploration with other third-party software. Specifically, these tools provide:

  • QC metrics and expression of housekeeping genes to qualify data
  • Hierarchical clustering of samples with user-defined gene sets or by all genes on the panel
  • Visualization with PCA, heatmaps, and gene expression distribution plots
  • Sample correlation using rich correlation plots
  • Fold-change estimates in easy-to-use tabular data format (TAC)
  • Volcano plots to visualize differentially genes (TAC)
Specifications
For Use With (Application)Oncology and Immune Therapy Gene Expression Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
No. of SamplesUp to 32 Samples
Nucleic Acid TypeRNA
PrimerAmpliSeq Primers
Product LineOncomine
Product TypeAssay
Quantity32 Samples
Sample TypeFFPE Tissue RNA
ThroughputUp to 4 Samples per Ion 520 Chip, 8 Samples per Ion 530 Chip
Workflow StepPanel and Library
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.

Frequently asked questions (FAQs)

With the Oncomine Immune Response Research Assay, how many samples can I multiplex per run?

Multiplexing can be done with 1-4 samples on the PGM 318 or S5/S5XL 520 chips, or with 2-8 samples on the S5/S5XL 520 chip. This enables flexibility for small-to-large retrospective profiling studies.

Which RNA isolation method do you recommend for use with the Oncomine Immune Response Research Assay?

We recommend using the RecoverAll Total Nucleic Acid Isolation Kit for FFPE (Cat. No. AM1975).

Can I automate the Oncomine Immune Response Research Assay workflow?

Yes, automated library and template preparation are enabled with the Ion Chef System.

Where can I find the User Guide for the Oncomine Immune Response Research Assay?

The User Guide can be found here (https://tools.thermofisher.com/content/sfs/manuals/MAN0015867_OncomineImmuneRespnseResrchAssay_UG.pdf).

What functional annotation groups are covered by the Oncomine Immune Response Research Assay?

The Oncomine Immune Response Research Assay content includes 395 genes across 36 functional annotation groups. These groups comprise genes associated with lymphocyte regulation, cytokine signaling, lymphocyte markers, checkpoint pathways, and tumor characterization.