The Ion GeneStudio S5 System is a semiconductor-based next-generation sequencing (NGS) system that enables simple targeted sequencing workflows. It is optimized to provide affordability for labs focused on smaller panels and lower sequencing throughput requirements. Combining an extended warranty with your system purchase helps extend the life of your investment.
This package includes the Ion GeneStudio S5 System, SmartStart orientation, and an extended warranty. Key features include:
• Select the chip (Ion 510–540 chip) that fits your application and throughput needs to achieve from 2–80 million reads per run
• Reduce setup time and complexity with cartridge-based plug-and-play reagents
• Complete run and analysis in hours and move forward with your experiments
• Increase efficiency and become operational faster with SmartStart orientation
• Receive fast repair from certified service engineers
• Minimize instrument downtime by troubleshooting with experienced Technical Support teams
For detailed instrument information and specifications, see the
Ion GeneStudio S5 System.
Get up and running quickly with SmartStartSmartStart orientation is carried out by a Field Application Scientist (FAS) to get you up and running quickly in your lab and reduce downtime. SmartStart is the first step of your education journey and is designed to help you work smarter and with greater efficiency. It includes:
• 1-day technology overview and instrument workflow
• 1-day hands-on application workflow training
• 10 hours bioinformatics consultancy
Content will vary depending on your level of knowledge and experience.
Extended warranty planAdd coverage to your existing manufacturer’s limited warranty with an extended warranty. Each extended warranty includes a planned maintenance visit. Should your instrument require a repair, the extended warranty coverage includes parts, labor, and travel at no additional cost. In addition, you’ll receive:
• On-site guaranteed 2-day response time*
• Scheduled on-site planned maintenance (PM)
• Priority access to Remote Service Engineers
Certain exclusions apply. Contact your Services and Support Representative for details at
service.sales@thermofisher.com.
*Availability limited in some geographical areas.
For Research Use Only. Not for use in diagnostic procedures.