Collibri™ Library Quantification Kit
Invitrogen™

Collibri™ Library Quantification Kit

The Invitrogen Collibri Library Quantification Kit includes a ready-to-use master mix optimized for Illumina NGS library quantification and a libraryRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A38524100100 Reactions
A38524500500 Reactions
Catalog number A38524100
Price (USD)
146.65
Online exclusive
165.00
Save 18.35
Each
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (USD)
146.65
Online exclusive
165.00
Save 18.35
Each
The Invitrogen Collibri Library Quantification Kit includes a ready-to-use master mix optimized for Illumina NGS library quantification and a library dilution buffer. Collibri Library Quantification Master Mix includes Platinum II Taq Hot Start DNA Polymerase and enables convenient and accurate quantification of NGS libraries. The master mix contains all components needed for the amplification reaction, including primers and passive reference dye, and does not require any preparation before use. Due to the universal concentration of the passive reference dye in the master mix, the kit can be used with any qPCR instrument without adjustments.

Features of the Collibri Library Quantification Kit include:
• Accurate quantification of libraries made from various sample types, including degraded samples
• Ease of use with all components ready to use—no reagent preparation required
• Visual cues—specially formulated dyes included to track pipetting steps
• Platinum hot-start technology—superior specificity, sensitivity, and room-temperature reaction setup

Unmatched convenience
Collibri Library Quantification Master Mix contains Platinum II Taq Hot Start DNA Polymerase, optimized buffer, dNTPs, primer mix targeting adapter sequences, SYBR green dye, passive reference dye, and a blue dye to track pipetting steps. This unique formulation provides ease of use at every step of the workflow:
• No reagent preparation
• Tracking of pipetting steps
• One cycling protocol for libraries of various lengths and GC/AT-content levels
• Universal passive reference dye concentration—a single master mix for all qPCR instruments

The Collibri Library Quantification Kit also includes a ready-to-use library dilution buffer that contains stabilizing agents to enable library integrity after dilution. The buffer contains yellow dye to track pipetting steps. When combined, the blue master mix and yellow diluted libraries become a green solution. This feature eases tracking of the addition of the diluted libraries or DNA standards to the master mix in the qPCR plate wells, helping reduce pipetting errors.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodSYBR
For Use With (Application)Illumina NGS Library Quantification
For Use With (Equipment)7500 Fast System, 7900HT Fast System, QuantStudio™, ViiA™ 7 System, StepOne™ System, StepOnePlus™ System
GC-Rich PCR PerformanceLow
No. of Reactions100 Reactions
PCR MethodqPCR
PolymerasePlatinum™ II Taq Hot Start DNA Polymerase
Product LineCollibri
Product TypeLibrary Quantification Kit
Quantity100 reactions
Sample TypeDNA, RNA
Shipping ConditionDry Ice
Test TimeStandard
FormatKit
Unit SizeEach
Contents & Storage
Store at -20°C.

Frequently asked questions (FAQs)

My qPCR instrument requires ROX. Which ROX dye do you recommend using with the Collibri Library Quantification Kit?

The Collibri Library Quantification Kit contains the Collibri Library Quantification Master Mix that already contains a universal concentration of ROX passive reference dye, so there is no need for additional adjustment with ROX dye regardless of the instrument you are working with.

Do I need to dilute the library prior to qPCR using the Collibri Library Quantification Kit?

Yes, the prepared library has to be diluted prior to running qPCR. Generally, it is recommended to run two dilution points, 1:10000 and 1:100000. For an established workflow where approximate library quantity is known, dilutions can be adjusted, but they need to fall within the range of the Collibri DNA Standards. Note that libraries should be diluted in the Collibri Library Dilution buffer that is included in the Collibri Library Quantification Kit. Dilution of the library stabilizes library molecules at low concentrations, producing accurate and reproducible results.

With the Collibri Library Quantification Kit, what effect do the visual cues for mixing have on library quantification results?

Colors in the Collibri Library Quantification Kit components have no effect on the universal passive reference dye, qPCR performance, or quantification results.

Can Collibri Library Quantification Kit be used with NGS libraries prepared for Ion Torrent sequencing?

No. However, the Collibri Library Quantification Kit contains the Collibri Library Quantification Master Mix that has premixed primers which are compatible with P5 and P7 adaptor sequences specific to the Illumina platform.

Citations & References (2)

Citations & References
Abstract
Predominance of the SARS-CoV-2 Lineage P.1 and Its Sublineage P.1.2 in Patients from the Metropolitan Region of Porto Alegre, Southern Brazil in March 2021.
Authors:Franceschi VB, Caldana GD, Perin C, Horn A, Peter C, Cybis GB, Ferrareze PAG, Rotta LN, Cadegiani FA, Zimerman RA, Thompson CE
Journal:Pathogens
PubMed ID:34451453
Almost a year after the COVID-19 pandemic had begun, new lineages (B.1.1.7, B.1.351, P.1, and B.1.617.2) associated with enhanced transmissibility, immunity evasion, and mortality were identified in the United Kingdom, South Africa, and Brazil. The previous most prevalent lineages in the state of Rio Grande do Sul (RS, Southern Brazil), ... More
High-resolution microbiome analysis enabled by linking of 16S rRNA gene sequences with adjacent genomic contexts.
Authors:Kapustina Ž, Medžiune J, Alzbutas G, Rokaitis I, Matjošaitis K, Mackevicius G, Žeimyte S, Karpus L, Lubys A
Journal:Microb Genom
PubMed ID:34473015
Sequence-based characterization of bacterial communities has long been a hostage of limitations of both 16S rRNA gene and whole metagenome sequencing. Neither approach is universally applicable, and the main efforts to resolve constraints have been devoted to improvement of computational prediction tools. Here, we present semi-targeted 16S rRNA sequencing (st16S-seq), ... More