CTS™ Immune Cell Serum Replacement (SR)
Gibco™

CTS™ Immune Cell Serum Replacement (SR)

CTS Immune Cell Serum Replacement (SR) is a defined xeno-free formulation designed to support in vitro expansion of human immune cells. It is added as a serum substitute to a basal cell culture medium such as CTS OpTmizer T Cell Expansion SFM or CTS AIM V Medium for the expansion of human immune cells such as T cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
A4702901250 mLBag - BioProcess Container (BPC)
A470290350 mLBag - BioProcess Container (BPC)
A259610150 mLBottle
A2596102500 mLBottle
A47029021 LBag - BioProcess Container (BPC)
Catalog number A4702901
Price (USD)
1,458.00
Each
Quantity:
250 mL
Format:
Bag - BioProcess Container (BPC)
Price (USD)
1,458.00
Each

CTS Immune Cell Serum Replacement (SR) is a defined xeno-free formulation designed to support in vitro expansion of human immune cells. It is added as a serum substitute to a basal cell culture medium such as CTS OpTmizer T Cell Expansion SFM or CTS AIM V Medium for the expansion of human immune cells such as T cells, dendritic cells, and macrophages.

Serum replacement
Human serum, a key component in many immune cell culture protocols, is expensive, inherently variable, and requires extensive safety testing prior to use when manufacturing a cGMP-compliant adoptive cell therapy product. CTS Immune Cell SR was developed to provide a serum-free, xeno-free complete medium that would minimize the supply and safety risks associated with human serum.

CTS Immune Cell SR is a defined and robust formulation that can replace supplementation with human serum in several different base media without sacrificing any performance of your cells. It eliminates the time and cost of qualifying human serum, enabling a seamless transition from research to clinical applications.

Immune cell qualified
CTS Immune Cell SR supports equivalent or improved results for multiple immune cell types (T cells, dendritic cells, macrophages), when used at similar concentration as human serum.

Closed workflow compatible
CTS Immune Cell SR is available in bottle and bioprocess container formats. The bioprocess container chamber is constructed from Aegis 5-14 film. Versatile connection options allow for integration into your closed process.

CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485-certified. Gibco CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.*

Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and access to our Drug Master File (DMF) are available.

*Other aspects of USP will be the responsibility of the end-user to assess. Thermo Fisher Scientific cannot fulfill USP in regards to application and therapy-specific aspects (e.g., residual assessment and removal of the Ancillary Material).

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell LineHuman Immune Cells
FormatBag - BioProcess Container (BPC)
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Product TypeSerum Replacement
Quantity250 mL
Shelf Life12 months from date of manufacture
Shipping ConditionDry Ice
ClassificationXeno-free
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Store flat at -5°C to -20°C.

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

What are the storage conditions for CTS Immune Cell Serum Replacament (SR)? Upon thawing, can I aliquot and refreeze at -20 degrees C?

After completely thawing the initial product, it can be aliquoted and refrozen for another usage. The thawed aliquots are stable for 4 weeks when stored at 2-8 degrees C.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.