ABgene Storage Plate, 96-well, 2.2 mL, individually wrapped, white

Catalog number: AB0661W

Thermo Scientific™  Related applications: Cell Counting, Viability, & Cryopreservation | Microplates, Dishes and Flasks

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific ABgene 2.2 mL white polypropylene plates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily.

Features of Thermo Scientific 2.2 mL storage and assay plates

• Designed for optimal sample recovery
• White color are ideal for luminescence-based assays.
• Each square well is specifically designed with an independent sealing rim to prevent cross-contamination
• U-bottom to aid resuspension

All plates are manufactured in our cleanroom facility to ensure molecular grade quality. All plates are ANSI format for compatibility with automated systems. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: 96-well plate
Plate Type: Storage Plate
Product Size: 50 plates
Shipping Condition: ELMS

Documents

Manuals & protocols