Poly(A) Tailing Kit with Manual
Catalog number: AM1350M
Invitrogen™  Related applications: In Vitro Transcription | Protein Expression
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
For the polyadenylation of in vitro transcribed RNA to enhance translation initiation efficiency. Sufficient reagents are included for 25 reactions.
• Adds a poly(A) tail of at least 150 nucleotides in length to the 3' termini of RNA
• Enhances translational efficiency in vivo
• Reaction can be adjusted to control for tail length
• Optimized for use with RNA transcripts synthesized using the mMESSAGE mMACHINE™ Kit
The Poly(A) Tailing Kit uses E. coli Poly(A) Polymerase I (E-PAP) to polyadenylate the 3'-termini of in vitro transcribed RNA. Polyadenylation plays an important role in the stabilization of RNA in eukaryotes and enhances the efficiency of translation initiation. In the Poly(A) Tailing Kit, the E-PAP reaction has been optimized so that mRNAs are efficiently tailed with at least 150 adenines. The additional adenine residues confer stability to the mRNA and may increase translational efficiency of in vitro synthesized capped RNA in microinjection and transfection experiments (see Figure).
Accessory Products:
The Poly(A) Tailing Kit is optimized for use with Ambion's mMESSAGE mMACHINE™ High Yield Capped RNA Transcription Kit. mMESSAGE mMACHINE™ Kits are available with T7 (SKU #AM1334), T3 (SKU #AM1338), or SP6 (SKU #AM1330) polymerases.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Starting Material: RNA (In Vitro Transcribed)
Final Product: In Vitro Transcribed RNA (Polyadenylated)
Product Line: Ambion™
Product Size: 25 reactions
Contents & storage
E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, and Control DNA Template should be stored at –20°C. Nuclease-free Water may be stored at any temperature.
Documents
Manuals & protocols