Thermo Scientific™
PTFE-Lined Closed Caps
Catalog number: B267-2414UB
Have Questions?
Thermo Scientific™
PTFE-Lined Closed Caps
Catalog number: B267-2414UB
Have Questions?
Thermo Scientific™ PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials. The same high quality PTFE-lined caps used on our Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.
Catalog Number
B267-2414UB
Unit Size
Each
dimClosureSize
Level
Unprocessed
No. per Case
144
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case144
Closure ColorBlue
LevelUnprocessed
Liner MaterialSilicone/PTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure Size24-414 mm
Unit SizeEach
Catalog Number
Unit Size
Case of 144
dimClosureSize
Level
Unprocessed
No. per Case
144
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case144
Closure ColorWhite
LevelUnprocessed
Liner MaterialSilicone/PTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size24-414 mm
Unit SizeCase of 144
Catalog Number
Unit Size
Case of 24
dimClosureSize
Level
Unprocessed
No. per Case
24
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size100-400 mm
Unit SizeCase of 24
Catalog Number
Unit Size
Case of 24
dimClosureSize
Level
Unprocessed
No. per Case
24
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
For Use With (Equipment)Thermo Scientific Glass Environmental Sample Bottles, Jars, Jugs and Vials
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size33-430 mm
Unit SizeCase of 24
Catalog Number
Unit Size
Each
dimClosureSize
Level
Unprocessed
No. per Case
24
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialSilicone/PTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeSolid Top
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size22-400 mm
Unit SizeEach
Catalog Number
Unit Size
Case of 24
dimClosureSize
Level
Unprocessed
No. per Case
24
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeSolid Top
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size38-430 mm
Unit SizeCase of 24
Catalog Number
Unit Size
Case of 24
dimClosureSize
Level
Unprocessed
No. per Case
24
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeSolid Top
For Use With (Equipment)Thermo Scientific Glass Environmental Sample Bottles, Jars, Jugs and Vials
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size45-400 mm
Unit SizeCase of 24
Catalog Number
Unit Size
Case of 24
dimClosureSize
Level
Unprocessed
No. per Case
24
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size89-400 mm
Unit SizeCase of 24
Catalog Number
Unit Size
Case of 24
dimClosureSize
Level
Unprocessed
No. per Case
24
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size28-400 mm
Unit SizeCase of 24
Catalog Number
Unit Size
Case of 24
dimClosureSize
Level
Unprocessed
No. per Case
24
Material
Polypropylene
Price (USD)
Availability
-
Full specifications
Quantity
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
For Use With (Equipment)Thermo Scientific Glass Environmental Sample Bottles, Jars, Jugs and Vials
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size33-430 mm
Unit SizeCase of 24
Showing 10 of 15Show more
Catalog NumberSpecificationsUnit SizedimClosureSizeLevelNo. per CaseMaterialPrice (USD)AvailabilityQuantity
B267-2414UBFull specifications
EachUnprocessed144Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case144
Closure ColorBlue
LevelUnprocessed
Liner MaterialSilicone/PTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure Size24-414 mm
Unit SizeEach
B267-2414UBBFull specifications
Case of 144Unprocessed144Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case144
Closure ColorWhite
LevelUnprocessed
Liner MaterialSilicone/PTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size24-414 mm
Unit SizeCase of 144
B167-1100BLFull specifications
Case of 24Unprocessed24Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size100-400 mm
Unit SizeCase of 24
B167-3330BLFull specifications
Case of 24Unprocessed24Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
For Use With (Equipment)Thermo Scientific Glass Environmental Sample Bottles, Jars, Jugs and Vials
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size33-430 mm
Unit SizeCase of 24
B267-2200UBFull specifications
EachUnprocessed24Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialSilicone/PTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeSolid Top
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size22-400 mm
Unit SizeEach
B167-3830BLFull specifications
Case of 24Unprocessed24Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeSolid Top
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size38-430 mm
Unit SizeCase of 24
B167-4500BLFull specifications
Case of 24Unprocessed24Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeSolid Top
For Use With (Equipment)Thermo Scientific Glass Environmental Sample Bottles, Jars, Jugs and Vials
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size45-400 mm
Unit SizeCase of 24
B167-8900BLFull specifications
Case of 24Unprocessed24Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size89-400 mm
Unit SizeCase of 24
B167-2800BLFull specifications
Case of 24Unprocessed24Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size28-400 mm
Unit SizeCase of 24
B167-3300BLFull specifications
Case of 24Unprocessed24Polypropylene
-
Certification ParametersNone
ColorWhite
DescriptionClosed top cap
MaterialPolypropylene
No. per Case24
Closure ColorWhite
LevelUnprocessed
Liner MaterialPTFE
Certifications/ComplianceManufactured under an ISO certified quality management system.
Closure IncludedYes
ShapeRound
Product TypeClosure with Bonded Liner
For Use With (Equipment)Thermo Scientific Glass Environmental Sample Bottles, Jars, Jugs and Vials
Closure LinerPTFE
Closure MaterialPolypropylene
Closure Size33-430 mm
Unit SizeCase of 24
Showing 10 of 15Show more
  • White polypropylene closed top cap with bonded PTFE liner
  • 24-414 cap has a PTFE-faced silicone liner for added cushion and an extra secure seal when used with bottles and VOA vials
Includes:
Cap with bonded liner
Compatible with:
For use with Thermo Scientific glass environmental sample bottles, jars, jugs and vials.