Phalloidin Labeling Probes
Invitrogen™

Phalloidin Labeling Probes

Achieve precise and reliable F-actin staining with fluorescent and biotinylated phalloidins. Phalloidin conjugates are widely used in imaging applications to selectively label F-actin in a variety of sample types including fixed and permeabilized cells, tissue sections, and cell-free experiments.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColorIntervalo de longitud de onda de excitaciónTipo de colorante
B7474NingunoNingunoBiotin-XX
A22281Azul346⁄442Alexa Fluor™ 350
A30104Violeta405/450Alexa Fluor™ Plus 405
A12379Verde495⁄518Alexa Fluor™ 488
O7466Verde496⁄520Oregon Green™ 488
F432Verde496⁄516FITC (fluoresceína)
A22282Amarillo531⁄554Alexa Fluor™ 532
R415Rojo anaranjado540⁄565TRITC (isotiocianato de tetrametilrodamina)
A22283Naranja556⁄570Alexa Fluor™ 546
A34055Naranja555⁄565Alexa Fluor™ 555
A30106Naranja555/565 nmAlexa Fluor Plus 555
B3475Rojo558⁄569BODIPY™
A12380Rojo anaranjado578⁄600Alexa Fluor™ 568
A12381Rojo581⁄609Alexa Fluor™ 594
T7471Rojo591⁄608Texas Red™
A22284Rojo lejano632⁄647Alexa Fluor™ 633
A34054Rojo lejano633⁄647Alexa Fluor™ 635
A22287Rojo lejano650⁄668Alexa Fluor™ 647
A30107Rojo lejano650/668 nmAlexa Fluor Plus 647
A22285Infrarrojo cercano663⁄690Alexa Fluor™ 660
A22286Infrarrojo cercano679⁄702Alexa Fluor™ 680
A30105Infrarrojo cercano758/784Alexa Fluor™ Plus 750
P3457NingunoNingunoFaloidina (sin marcar)
Número de catálogo B7474
Precio (MXN)
-
Color:
Ninguno
Intervalo de longitud de onda de excitación:
Ninguno
Tipo de colorante:
Biotin-XX
Fluorescent and biotinylated phalloidins are water soluble and bind to filamentous actin (F-actin) with nanomolar affinity, making them convenient probes for labeling, identifying, and quantifying F-actin in cryopreserved tissue sections, fixed and permeabilized cells, and cell-free experiments. Phalloidin conjugates bind similarly to actin from various species, including plants and animals, enabling staining of the cytoskeleton in a wide range of samples.

A variety of phalloidin conjugates for filamentous (F-actin) staining are available, including fluorescent Alexa Fluor and Alexa Fluor Plus phalloidins, along with phalloidins conjugated to classic fluorescent dyes such as BODIPY, fluorescein, and rhodamine. Phalloidin staining is spectrally compatible with other fluorescent stains used in cellular analyses such as GFP/RFP, Qdot nanocrystals, and other Alexa Fluor conjugates and antibodies. Biotin‐XX Phalloidin can be used to visualize actin filaments via fluorescent streptavidin tags or standard enzyme-mediated avidin/streptavidin techniques such as in electron microscopy. Unlabeled phalloidin is available for use as a control in blocking F‐actin staining or in promoting polymerization.

Phalloidin conjugates bind to both large and small actin filaments with similar affinity in a 1:1 stoichiometry between phallotoxin and actin subunits. They do not bind G-actin monomers.

Alexa Fluor and Alexa Fluor Plus phalloidin conjugates for F-actin staining

Fluorescent Alexa Fluor dye conjugates of phalloidin are popular F-actin stains, offering color choices across the full spectral range. These phalloidin conjugates provide researchers with fluorescent probes that are superior in brightness and photostability compared to other spectrally similar conjugates.

Alexa Fluor Plus Phalloidin conjugates retain the same specificity for actin but offer 3-5 times greater sensitivity and brightness compared to the corresponding Alexa Fluor Phalloidin conjugate. This increased brightness is beneficial for challenging F-actin imaging, such as the super‐resolution microscopy methods SIM and STORM, and for reliable staining of actin stress fibers.

Features of phalloidin probes

  • High specificity—binds selectively to F-actin, which allows for precise labeling of actin filaments in fixed cells and cryopreserved tissues
  • Strong affinity—nanomolar binding affinity for F-actin, which ensures stable and reliable actin staining
  • Extensive fluorescent conjugate options—over twenty conjugated varieties of phalloidin
  • Compatibility with fixed samples—typically used with fixed cells and tissues, making them suitable for actin staining in detailed structural studies, immunofluorescence staining, and IHC applications
  • Multiplexing capability—the wide availability of phalloidin conjugates enables their use in combination with other fluorescent probes and antibodies for multiplex imaging. Biotinylated phalloidin can be made use of in downstream streptavidin steps.
  • Quantitative analysis—can be used for quantitative analysis of F-actin distribution and density within cells, aiding in the study of cytoskeletal dynamics. The unlabeled phalloidin can be titrated as a control.
  • Ease of use—staining is straightforward and quick
  • Excellent stability—exhibit good photostability, which is essential for prolonged imaging sessions and time-lapse studies
  • Wide applicability—used for a range of applications, including studying cell morphology, motility, and the effects of drugs on the actin cytoskeleton

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
ColorNinguno
Tipo de coloranteBiotin-XX
Intervalo de longitud de onda de excitaciónNinguno
Para utilizar con (equipo)Fluorescence Microscope, Flow Cytometer, Confocal Microscope
Cantidad50 Units
Condiciones de envíoTemperatura ambiente
Tipo de etiquetaBiotina y otros haptenos
Tipo de productoFaloidina
Sub Cellular LocalizationActina, citoesqueleto, Cytoskeleton
Unit SizeEach
Contenido y almacenamiento
Almacenar en congelador entre -5 °C y -30 °C

Preguntas frecuentes

I want to label F-actin with phalloidin and image with transmission electron microscopy. What is the best way to go about it?

One way is to use a biotin-phalloidin conjugate, then label that with a colloidal gold conjugate of streptavidin. You will need to block endogenous biotin first. Another method is to label with fluorescein phalloidin, then with an anti-fluorescein antibody conjugated with colloidal gold.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Do you offer alternative package sizes for phalloidin or ReadyProbes reagents?

Alternative package sizes may be available through a custom request process. Contact Thermo Fisher Scientific customer support or your sales representative to discuss available options for the specific reagent and quantity needed.

Can I use conjugated phalloidin label probes on organoids?

Yes. Conjugated phalloidin labeling probes can be used with organoids when F-actin is accessible after fixation and permeabilization. Because organoids are thicker than monolayer cell cultures, protocol optimization may be needed, including permeabilization conditions, probe incubation time, and probe concentration.

Are conjugated phalloidin reagents antibodies?

No, Phalloidin is not an antibody. It is a bicyclic peptide toxin originally isolated from the Amanita phalloides mushroom and is used to bind and label F-actin in fixed and permeabilized samples.

Can I use methanol instead of DMSO to reconstitute the conjugated phalloidins from the lyophilized solids?

Yes, methanol can be used to reconstitute fluorescent phalloidin, but it creates a less concentrated stock solution than DMSO. Dissolve the vial contents in 1.5 mL of methanol to yield a 40X stock solution at a concentration of 200 assays/mL, which is equivalent to approximately 6.6 µM. One unit/assay of fluorescent phalloidins is equivalent to 5 µL of the methanolic stock solution. Methanol may be used as a stock-solution solvent, but methanol-containing fixatives are not recommended because they can disrupt F-actin.

Citations & References (22)

Citations & References
Abstract
Expanding the multicolor capabilities of basic confocal microscopes by employing red and near-infrared quantum dot conjugates.
Authors:Kingeter LM, Schaefer BC,
Journal:BMC Biotechnol
PubMed ID:19463154
'BACKGROUND: Confocal microscopy is a widely employed methodology in cellular biology, commonly used for investigating biological organization at the cellular and sub-cellular level. Most basic confocal microscopes are equipped to cleanly discriminate no more than four fluorophores in a given sample, limiting the utility of this method for co-localization, co-expression, ... More
Wet electron microscopy with quantum dots.
Authors:Timp W, Watson N, Sabban A, Zik O, Matsudaira P
Journal:Biotechniques
PubMed ID:16989089
Wet electron microscopy (EM) is a new imaging method with the potential to allow higher spatial resolution of samples. In contrast to most EM methods, it requires little time to perform and does not require complicated equipment or difficult steps. We used this method on a common murine macrophage cell ... More
Phallotoxin and actin binding assay by fluorescence enhancement.
Authors:Huang ZJ, Haugland RP, You WM, Haugland RP
Journal:Anal Biochem
PubMed ID:1595896
The fluorescence of five fluorophores conjugated to phallotoxins was found to be specifically enhanced upon binding to F-actin in a polymerizing buffer. Rhodamine phalloidin had the greatest fluorescence enhancement of ninefold. The fluorescence titration of rhodamine phalloidin by actin was shown to be consistent with stoichiometric binding. The fluorescence enhancement ... More
Activity-dependent tau protein translocation to excitatory synapse is disrupted by exposure to amyloid-Beta oligomers.
Authors:Frandemiche ML, De Seranno S, Rush T, Borel E, Elie A, Arnal I, Lanté F, Buisson A,
Journal:
PubMed ID:24760868
Tau is a microtubule-associated protein well known for its stabilization of microtubules in axons. Recently, it has emerged that tau participates in synaptic function as part of the molecular pathway leading to amyloid-beta (Aß)-driven synaptotoxicity in the context of Alzheimer's disease. Here, we report the implication of tau in the ... More
Detection of small differences in actomyosin function using actin labeled with different phalloidin conjugates.
Authors:Balaz M, Månsson A
Journal:Anal Biochem
PubMed ID:15745742
This study shows that there is only a negligible difference in actomyosin function in the in vitro motility assay among actin filaments labeled with Rhodamine phalloidin (RhPh), Alexa-488 phalloidin (APh), and biotin-XX phalloidin (BPh). Similar results were obtained at varying ionic strengths (0.02-0.13 M), in the presence of imidazole or ... More