Orbitrap Exploris™ MX Mass Detector
Orbitrap Exploris™ MX Mass Detector
Orbitrap Exploris™ MX Mass Detector
Orbitrap Exploris™ MX Mass Detector
Orbitrap Exploris™ MX Mass Detector
Thermo Scientific™

Orbitrap Exploris™ MX Mass Detector

Monitor product quality attributes in the Late Stage Drug Development and the critical quality attributes in Manufacturing for lot release with Thermo Scientific™ Orbitrap Exploris™ MX mass detector.
Have Questions?
Catalog NumberType
BRE725536Orbitrap LC-MS
Catalog number BRE725536
Price (USD)
-
Type:
Orbitrap LC-MS

Achieve routine mass monitoring in your biopharmaceutical laboratory with the fit-for-purpose Thermo Scientific™ Orbitrap Exploris™ MX mass detector. This system is simple to operate and compliance ready with Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS). HRAM data provides the utmost confidence for sequence confirmation, peptide monitoring, intact protein analysis, glycan profiling, as well as oligonucleotides and their impurity mass confirmation.

Simplicity for scientists and technicians
Chromeleon CDS offers built in methods, control, data processing and management that simplify tasks and reduce errors for more 'right first time' analyses. A suite of smart tools make work faster and easier, while ensuring reproducible, high-quality mass monitoring.

Productivity to accelerate commercialization
High uptime boosts productivity. Rapid 'set and forget' calibration procedures provide consistent mass stability for at least four weeks at prescribed conditions. Instrument status monitoring and optimized pressure control alert users when maintenance is needed, avoiding unnecessary downtime and repeat analyses.

Compliance for competitive advantage
Designed for data integrity, data security, and compliance, fully scalable Chromeleon CDS gives you a competitive edge in meeting global regulatory requirements.

Consistent results deliver confidence
Robust and reliable performance from system to system and from site to site ensure high-confidence results used to make critical decisions. Industry-proven Orbitrap technology, now purposely designed for mass monitoring in future-proof lab operations.

Seamless method transfer streamlines multi-attribute method (MAM)
Easy transfer of sophisticated MAM assays from development to manufacturing bridges the gap between development and QC.

Dedicated global support maximizes your productivity
Our dedicated MAM team maximizes your productivity and confidence with installation, training, and fast service and support. Standardized system performance evaluation test (SET) and installation qualification/operational qualification (IQ/OQ) thoroughly assess system performance with comprehensive installation and post-maintenance acceptance criteria.

Thermo Fisher Scientific is working to provide more information about our products' environmental impact by stepping up to participate in the ACT Label program with selected products.

The Thermo Scientific Orbitrap Exploris and Excedion series mass spectrometers are Orbitrap instruments with an atmospheric pressure ionization (API) source for liquid chromatography (LC) mass spectrometry (MS) high-throughput applications. The instruments are designed to be placed on a bench in the laboratory and maintain mass accuracy across chromatographic peaks, supporting confident identification and quantitation using high-resolution accurate-mass detection.

Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.

Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.

For General Lab Use Only. Not For Diagnostic Procedures.

Specifications
ACT LabelYes
DescriptionOrbitrap Exploris MX mass detector
Dynamic Range>5000 within a single Orbitrap mass analyzer spectrum
Mass AccuracyExternal calibration achieves <3 ppm RMS drift over 24 hours, 1-point calibration achieves <3 ppm RMS drift over at least 4 weeks, Internal lock mass calibration achieves <1 ppm RMS drift over 24 hours, EASY-IC achieves <1 ppm RMS drift for at least 5 days
ModelOrbitrap Exploris MX mass detector
Polarity SwitchingOne Full Scan cycle ** <700 ms equals> 1.4 Hz
Scan RatesUp to 22 Hz at resolution setting 15,000
Weight (Metric)120 kg (265 lb.) without data system, vacuum rough pumps, and optional items
Mass Range40 to 3000 m/z, Up to 8000m/z with BioPharma Option
Max. ResolutionUp to 180,000 at 200 m/z
TypeOrbitrap LC-MS
Unit SizeEach