CellEvent™ Caspase-3/7 Green Flow Cytometry Assay Kit
Invitrogen™

CellEvent™ Caspase-3/7 Green Flow Cytometry Assay Kit

The CellEvent™ Caspase-3/7 Green Flow Cytometry Assay Kit enables the flow cytometric detection of activated caspase-3 and caspase-7 in apoptotic자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
C1074020 Assays
C10427100 Assays
카탈로그 번호 C10740
제품 가격(KRW)
185,000
온라인 행사
Ends: 30-Jun-2026
217,000
할인액 32,000 (15%)
Each
수량:
20 Assays
제품 가격(KRW)
185,000
온라인 행사
Ends: 30-Jun-2026
217,000
할인액 32,000 (15%)
Each

The CellEvent™ Caspase-3/7 Green Flow Cytometry Assay Kit enables the flow cytometric detection of activated caspase-3 and caspase-7 in apoptotic cells. The kit includes the novel fluorogenic substrate CellEvent™ Caspase-3/7 Green Detection Reagent, as well as SYTOX™ AADvanced™ Dead Cell Stain.

• Caspase-3/7 specific—includes the recognition sequence for activated caspase-3 and caspase-7
• Easy identification—clearly identify live, dead, and apoptotic cell populations
• Quick analysis—washing and fixation is not required
• Multicolor compatibility—combine with other dyes excitable by the 488 nm laser or other lasers

CellEvent™ Caspase-3/7 Green Detection Reagent is a cell-permeant reagent that consists of a four-amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye. During apoptosis, caspase-3 and caspase-7 proteins are activated and able to cleave the caspase 3/7 recognition sequence encoded in the DEVD peptide. Cleavage of the recognition sequence and binding of DNA by the reagent labels the apoptotic cells with a bright, fluorogenic signal that has absorption/emission maxima of ∼511/533 nm. When used together with the SYTOX™ AADvanced™ Dead Cell Stain, apoptotic cells can be easily discriminated from live and necrotic cells.

Because no single parameter defines apoptosis in all systems, we strongly suggest using a combination of different measurements for reliable detection of apoptosis.

For Research Use Only. Not for use in diagnostic procedures.
사양
여기/방출SYTOX AADvanced: 546/647
CellEvent Caspase 3/7 Green Detection Reagent: 511/533
유세포분석기 레이저 라인488
용도(애플리케이션)Flow Cytometry
용도(장비)Attune™ Acoustic Focusing Cytometer, Flow Cytometer
라벨 유형Other Label(s) or Dye(s)
라벨 또는 염료CellEvent™ Caspase 3/7 Green Detection Reagent, SYTOX AADvanced dead cell stain
제품라인CellEvent
제품 유형Caspase Assay Kit
수량20 Assays
배송 조건Wet Ice
검출 방법Fluorescence
형식Kit
Unit SizeEach
구성 및 보관
Contains 1 vial CellEvent™ Caspase 3/7 Green Detection Reagent, 1 vial SYTOX AADvanced dead cell stain, and 1 vial of DMSO.

Store all vials in the kit upright, at or below -20deg;C, desiccated, and protected from light.

자주 묻는 질문(FAQ)

Can I use CellEvent Caspase-3/7 Green Detection Reagent with fixed samples?

No, it must be applied to live cell populations.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

With the CellEvent Caspase-3/7 Green Detection Reagent, may I stain cells with nucleic acid stains?

Yes, you may use SYTOX AADvanced Dead Cell Stain that is included in the CellEvent Caspase-3/7 Green Ready Flow Reagent (Cat. No. R37167) and in the CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit (Cat. No. C10427, C10740). You may also try using other nucleic acid stains in the red or far red channels for staining cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

With the CellEvent Caspase-3/7 Green Detection Reagent, may I multiplex with fluorescent immunolabeling?

Yes. You can apply surface labels or other reagents. We recommend applying the surface labels or other reagents first.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is the CellEvent Caspase-3/7 Green Detection Reagent fixable?

The CellEvent Caspase-3/7 Green Detection Reagent is not covalently attached to any cellular component and may be lost upon fixation. For flow cytometry applications, after applying the reagent, you should examine without washing or fixation. For imaging applications, the CellEvent product signal may be retained after fixation in 3.7% formaldehdye for 15 min.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I fix my CellEvent Caspase 3/7 - labeled cells?

For imaging applications, we recommend fixing with 3.7% formaldehyde at room temperature for 15 mins. After fixation, cells can be imaged for up to 24 hrs, but the signal will gradually degrade over time. For flow cytometry applications, fixation is not recommended.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

인용 및 참조 문헌 (7)

인용 및 참조 문헌
Abstract
Clumping and Viability of Bone Marrow Derived Mesenchymal Stromal Cells under Different Preparation Procedures: A Flow Cytometry-Based In Vitro Study.
Authors:Cui LL, Kinnunen T, Boltze J, Nystedt J, Jolkkonen J
Journal:Stem Cells Int
PubMed ID:27022399
'Complications of microocclusions have been reported after intra-arterial delivery of mesenchymal stromal cells. Hence, quantification and efficient limitation of cell clumps in suspension before transplantation is important to reduce the risk. We used a flow cytometry-based pulse-width assay to assess the effects of different cell suspension concentrations (0.2-2.0 × 10(6)/mL), ... More
New flow cytometry approaches in equine andrology.
Authors:Peña FJ, Ortega Ferrusola C, Martín Muñoz P
Journal:Theriogenology
PubMed ID:27160445
Flow cytometry is currently recognized as a robust tool for the evaluation of sperm quality and function. However, within equine reproduction, this technique has not reached the sophistication of other areas of biology and medicine. In recent years, more sophisticated flow cytometers have been introduced in andrology laboratories, and the ... More
Avenanthramide-C reduces the viability of MDA-MB-231 breast cancer cells through an apoptotic mechanism.
Authors:Hastings J, Kenealey J
Journal:Cancer Cell Int
PubMed ID:29075150
Avenanthramides (AVN) are a relatively unstudied family of phytochemicals that could be novel chemotherapeutics. These compounds, found in oats, are non-toxic to healthy cells and have been shown to reduce viability of human colon and liver cancers in vitro. However, these studies do not elucidate a molecular mechanism for individual ... More
Comparison of
Authors:Opydo-Chanek M, Sladowska K, Blicharski K, Mikeš J, Fedorocko P, Niemeyer U, Mazur L
Journal:Anticancer Res
PubMed ID:29061820
The oxazaphosphorines, ifosfamide and cyclophosphamide, represent a class of alkylating agents. The aim of the present in vitro study was to compare antileukemic activity of 4-hydroperoxyifosfamide (4-OOH-IF) and 4-hydroperoxycyclophosphamide (4-OOH-CP). ... More
Liposomal encapsulation of silver nanoparticles enhances cytotoxicity and causes induction of reactive oxygen species-independent apoptosis.
Authors:Yusuf A, Brophy A, Gorey B, Casey A
Journal:J Appl Toxicol
PubMed ID:29181855
Silver nanoparticles (AgNP) are one of the most widely investigated metallic NPs due to their promising antibacterial activities. In recent years, AgNP research has shifted beyond antimicrobial use to potential applications in the medical arena. This shift coupled with the extensive commercial applications of AgNP will further increase human exposure ... More