Concanavalin A (Con A) Conjugates
Invitrogen™

Concanavalin A (Con A) Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugateColorExcitation/EmissionProtein Subtype
C860TetramethylrhodamineOrange555/580 nmConcanavalin A (Con A)
C11254Alexa Fluor 350Blue346/442 nmConcanavalin A (Con A)
C11252Alexa Fluor 488Green495/519 nmConcanavalin A (Con A)
C21401Alexa Fluor 488Green495/519 nmConcanavalin A, succinylated
C11253Alexa Fluor 594Red590/617 nmConcanavalin A (Con A)
C21402Alexa Fluor 633Deep Red632/647 nmConcanavalin A (Con A)
C21421Alexa Fluor 647Deep Red650/668 nmConcanavalin A (Con A)
C56126Alexa Fluor Plus 405Violet405/450 nmConcanavalin A (Con A)
C56127Alexa Fluor Plus 750Near-infrared750/777 nmConcanavalin A (Con A)
C825Texas RedRed595/615 nmConcanavalin A (Con A)
Catalog number C860
Price (USD)
242.65
Online exclusive
264.00
Save 21.35
Each
Conjugate:
Tetramethylrhodamine
Color:
Orange
Excitation/Emission:
555/580 nm
Protein Subtype:
Concanavalin A (Con A)
Price (USD)
242.65
Online exclusive
264.00
Save 21.35
Each
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC). 

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange
Cross Reactivityα-man, α-glc
Excitation/Emission555/580 nm
Expression SystemNatural
Label TypeClassic Dyes
Ligand TypeLectin
Protein FamilyLectins
Protein SubtypeConcanavalin A (Con A)
Protein TagNone
Purity or Quality GradeImaging Quality
Quantity10 mg
Research CategoryImaging
Shipping ConditionRoom Temperature
SourceCanavlia ensiformis
ConjugateTetramethylrhodamine
For Use With (Application)Immunocytochemistry, Immunohistochemistry, Immunofluorescence
FormLyophilized
RecombinantNative
SpeciesC. ensiformis
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (56)

Citations & References
Abstract
Effects of the regulatory light chain phosphorylation of myosin II on mitosis and cytokinesis of mammalian cells.
Authors: Komatsu S; Yano T; Shibata M; Tuft R A; Ikebe M;
Journal:J Biol Chem
PubMed ID:10944522
'Myosin plays an important role in mitosis, especially during cytokinesis. Although it has been assumed that phosphorylation of regulatory light chain of myosin (RLC) controls motility of mammalian non-muscle cells, the functional significance of RLC phosphorylation remains uninvestigated. To address this problem, we have produced unphosphorylatable RLC (T18A/S19A RLC) and ... More
Spontaneous and ligand-induced trafficking of CXC-chemokine receptor 4.
Authors:Tarasova NI, Stauber RH, Michejda CJ
Journal:J Biol Chem
PubMed ID:9632631
'A chimeric protein consisting of CXC-chemokine receptor 4 (CXCR4) and the green fluorescent protein (GFP) was used for studying receptor localization and trafficking in real time in stably transduced HeLa, U-937, CEM, and NIH/3T3 cells. CXCR4-GFP was fully active as a co-receptor in mediating human immunodeficiency virus (HIV) entry. Both ... More
Flow cytometric measurement of fluorescence resonance energy transfer on cell surfaces. Quantitative evaluation of the transfer efficiency on a cell-by-cell basis.
Authors:Trón L, Szöllósi J, Damjanovich S, Helliwell SH, Arndt-Jovin DJ, Jovin TM
Journal:Biophys J
PubMed ID:6428482
'A method has been developed for the determination of the efficiency (E) of the fluorescence resonance energy transfer between moieties on cell surfaces by use of a computer-controlled flow cytometer capable of dual wavelength excitation. The absolute value of E may be calculated on a single-cell basis. The analysis requires ... More
Proximity of lectin receptors on the cell surface measured by fluorescence energy transfer in a flow system.
Authors:Chan SS, Arndt-Jovin DJ, Jovin TM
Journal:J Histochem Cytochem
PubMed ID:374620
'Molecules of the lectin concanavalin A have been labeled separately with the fluorescein and rhodamine chromophores and jointly bound to the surface of transformed Friend erythroleukemia cells. The two dyes constitute an ideal donor-acceptor pair for fluorescence resonance energy transfer thereby permitting the determination of the proximity relationships between bound ... More
Immobilization of concanavalin A receptors during differentiation of neuroblastoma cells.
Authors:Fishman MC, Dragsten PR, Spector I
Journal:Nature
PubMed ID:6261153
'Neuroblastoma cells serve as a useful model of neuronal development because compounds such as dimethyl sulphoxide (DMSO) and dibutyryl cyclic AMP cause them to undergo a process of controlled differentiation in tissue culture, during which they can extend long processes, develop characteristic excitability mechanisms, synthesize neurotransmitters and form synapses. We ... More