Automatically process multiple electronic personal dosimeters (EDPs) with the Thermo Scientific™ DI-10C Calibration Check Irradiator. The DI-10C is a complete irradiator system that provides separate beta and gamma calibration checks for Thermo Scientific™ MK2 Electronic Personal Dosimeters and Thermo Scientific™ EPD TruDose Electronic Personal Dosimeters. With an intuitive, easy to use control system, it is a very capable, but simple, system to use. A carousel holding up to 10 EPDs is loaded in the DI-10C irradiator. The system then automatically processes all EDPs and displays the results.
- Multiple verification protocols:
- Check against calibrated true values
- Check against reference EPD
- Check against previous measurements for the same EPD
- Measures and verifies doses and EPD channel counts
- Compact, bench top unit, fully self-contained
- No need for any additional shielding
- Accepts 10 EPDs in one load
- Operates automatically under software control
- Comprehensive safety features
- β and γ sources check all EPD counting channels
- Interfaces with the EPD maintenance database (EMDS4)
- Stand alone mode for irradiator calibration and/or machine diagnose