Thermo Scientific™
RA Lamb Seven-Drawer Filing Cabinets
 Related applications:
Anatomical Pathology
Store larger microscope slides or paraffin wax blocks in cardboard boxes in Thermo Scientific™ RA Lamb Seven-Drawer Filing Cabinets.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Store up to 1500 or 3000 76 x 51mm slides
  • Also store paraffin wax blocks in cardboard boxes, Super Mega Cassettes, photographic transparencies and most grid boxes used in electron microscopy
  • Use an RA Lamb divider to conveniently store two rows of 76 x 26mm slides
  • Choose from three colors to color-code large batches of slides and other laboratory equipment
Please Note: Availability may vary by country.
Documents
Manuals & protocols