Thermo Scientific™
RA Lamb BlockStore Storage Options for the LabStack Storage System
 Related applications:
Anatomical Pathology
Store lab supplies that don't typically fit in standard-size compartments with Thermo Scientific™ Ray A Lamb BlockStore Storage Options. The versatile, expandable system easily accommodates embedding rings, processing cassettes, and unmounted paraffin blocks. Dividers within each drawer further enhance the organization and accessibility of hard-to-store lab supplies, creating an efficient environment where processed tissue can be archived safely.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Compatibility:
  • Compatible with all standard LabStack cabinets and options
  • Accepts embedding rings, processing cassettes, and unmounted paraffin blocks
Includes:
  • 8-Drawers (arranged in two rows of four)
  • Enough dividers to produce seven rows, 1.2 in. (30mm) wide, in each drawer
Documents
Manuals & protocols