Hand-Held Magnetic Plate Washer
Invitrogen™
Hand-Held Magnetic Plate Washer
The Hand-Held Magnetic Plate Washer is designed for use with the ProcartaPlex assay configured for magnetic separation.
Have Questions?
Catalog NumberQuantity
EPX-55555-0001 Ea.
Catalog number EPX-55555-000
Price (HKD)
10,846.00
Each
Quantity:
1 Ea.
Price (HKD)
10,846.00
Each
The Hand-Held Magnetic Plate Washer is designed for use with the ProcartaPlex assay configured for magnetic separation. The handheld magnetic separator enables you to remove the liquid contents of the 96-well plate by simply decanting or flicking the contents into a sink and blotting off the remainder on a paper towel.
Please refer to the appropriate ProcartaPlex User Manual for details on running the assay.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypePlate Washer
Quantity1 Ea.
Unit SizeEach